Ohaus Explorer NTEP Approved Toploading Balances
The Explorer is perfect for university research, teaching laboratories, general lab and industrial applications
  • Intelligent performance – optimized linearity and repeatability
  • Intelligent calibration - AutoCal automatic internal calibration ensures your results are accurate
  • Intuitive operation – Large color touch-screen display and icon-driven application software
  • Intuitive software - SmarText™ 2.0 is OHAUS’ easy-to-use graphical software
  • Practical touchless sensors - Hands-free operation
  • Ingenious modular design - Display can be separated from the weighing base for maximum flexibility in any work area
The OHAUS Explorer’s advanced applications simplify even the most complex laboratory measurements. Whether it’s determining the difference between initial and residual weights or calculating the density of solids and liquids, Explorer eliminates the need for time consuming manual calculations and data logging. The high resolution display and innovative user interface make balance setup and application use effortless.
$8,284.87 - $11,094.63CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Ohaus EX10201N Explorer Toploading Balance, 10200g x 0.1g with NTEP Certified 1180051 Mfr # EX10201N
COMPARE
10200 100 to 240 0.1
$8,570.71CAD / Each
Ohaus EX10202N Explorer Toploading Balance, 10200g x 0.01g with NTEP Certified 1180052 Mfr # EX10202N
COMPARE
10200 100 to 240 0.01
$11,094.63CAD / Each
Ohaus EX4202N Explorer Toploading Balance, 4200g x 0.01g with NTEP Certified 1180054 Mfr # EX4202N
COMPARE
4200 100 to 240 0.01
$8,284.87CAD / Each