Ohaus® Explorer™ EXR Series NTEP Precision Toploading Balances
High-performance balances with NTEP certification offer intuitive interface and outstanding accuracy
  • Intuitive 7" full-color VGA touchscreen simplifies operation
  • Advanced weighing applications and fast stabilization improve productivity
  • User management and audit trail support GLP/GMP compliance
  • Flexible connectivity with USB, RS-232, and optional Ethernet
  • Full metal housing ensures durability in demanding environments
  • NTEP legal-for-trade and Measurement Canada approved for regulated weighing applications
Explorer™ EXR precision balances with NTEP certification are designed for regulated environments requiring legal-for-trade compliance without compromising performance. The intuitive 7" full-color VGA touchscreen simplifies operation, while fast stabilization and advanced weighing applications improve productivity in laboratory and industrial settings. Automatic, user-triggered, and span calibration options ensure precise measurements; the built-in automatic calibration performs self-calibration using an internal mass. Select from 11 application modes: Basic Weighing, Parts Counting, Check Counting, Percent Weighing, Check Weighing, Dynamic Weighing, Totalization, Formulation, Differential, Density Determination, Peak Hold.
Easily manage weighing operations on the 7" touchscreen display. An easy-to-clean, chemical-resistant glass panel displays vibrant colors for easy viewing. Touchless sensors located on the terminal activate often used functions such as print, zero, tare, or calibrate, simply wave your hand over the sensors to activate. Built-in RS-232, USB (A and C), and Ethernet connections provide seamless data transfer; order optional dongle for Bluetooth® and Wi-Fi capabilities. Save and recall up to 1000 sample library records to simplify your weighing operation. Each record may contain minimum weight, sample name, sample ID, batch ID, project name, and more.
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Repeatability (mg)
Availability Pricing
Ohaus EXR223N Explorer™ EXR Series - NTEP Precision Top Loading  Balance, 220g x 1mg, Draftshield and Touch Screen 2060550 Mfr # EXR223N
COMPARE
220 0.001 0.7
$6,117.52CAD / Each
Ohaus EXR423N Explorer™ EXR Series - NTEP Precision Top Loading  Balance, 420g x 1mg, Draftshield and Touch Screen 2060551 Mfr # EXR423N
COMPARE
420 0.001 0.7
$7,154.78CAD / Each
Ohaus EXR623N Explorer™ EXR Series - NTEP Precision Top Loading  Balance, 620g x 1mg, Draftshield and Touch Screen 2060552 Mfr # EXR623N
COMPARE
620 0.001 0.7
$8,741.62CAD / Each
Ohaus EXR1203N Explorer™ EXR Series NTEP Precision Top Loading  Balance, 1200g x 1mg, Draftshield and Touch Screen 2060553 Mfr # EXR1203N
COMPARE
1200 0.001 0.7
$10,245.95CAD / Each
Ohaus EXR2202N Explorer™ EXR Series NTEP Precision Top Loading  Balance, 2200g x 0.01g, Touch Screen 2060554 Mfr # EXR2202N
COMPARE
2200 0.01 7
$6,373.89CAD / Each
Ohaus EXR4202N Explorer™ EXR Series NTEP Precision Top Loading  Balance, 4200g x 0.01g, Touch Screen 2060555 Mfr # EXR4202N
COMPARE
4200 0.01 7
$7,324.22CAD / Each
Ohaus EXR6202N Explorer™ EXR Series NTEP Precision Top Loading  Balance, 6200g x 0.01g, Touch Screen 2060556 Mfr # EXR6202N
COMPARE
6200 0.01 7
$8,312.87CAD / Each
Ohaus EXR8202N Explorer™ EXR Series NTEP Precision Top Loading  Balance, 8200g x 0.01g, Touch Screen 2060557 Mfr # EXR8202N
COMPARE
8200 0.01 7
$9,081.98CAD / Each
Ohaus EXR12202N Explorer™ EXR Series NTEP Precision Top Loading  Balance, 12200g x 0.01g, Touch Screen 2060558 Mfr # EXR12202N
COMPARE
12200 0.01 7
$9,859.93CAD / Each
Showing 1- 9 of 9