Ohaus Bead Genie™ Vortex Shakers
Unique vertical motion, to ensure smooth and efficient mixing
  • Compact design fits on the lab bench, into a bio safety cabinet, or for use in your cold room
  • Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop
  • Variable “RUN/STOP” time provides rapid pulsing for superior interval mixing
  • Digital display provides accuracy, reproducibility, and repeatability with complete control of speed and time
Bead Genie™ vortex shakers provide easy and accurate control of speed and time with a digital display. The variable speed shaking allows you to adjust the level of mixing from gentle shaking to vigorous vortexing. Easy-to-change platforms allow you to adapt your shaker to a variety of applications. This shaker is ideal for lysing, extraction, or homogenization applications, utilizing an aggressive vertical mixing motion. Supplied with holders to hold vertically 12 microtubes (1.5 or 2.0 mL), 6 x 15 mL tubes, or 3 x 50 mL tubes, and 12 microtubes (1.5 or 2.0 mL) horizontally. Beads are also available and sold separately for complete bead beating solution.
$3,247.35 - $3,945.74CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
Ohaus Bead Genie™ Vortex Shaker, 250 to 2400 rpm; 120 VAC 5140203 Mfr # SI-B100
COMPARE
2400 120 Bead Genie™ Vortex Shaker, 250 to 2400 rpm; 120 VAC
$3,945.74CAD / Each
Ohaus Bead Genie™ Vortex Shaker, 250 to 2400 rpm, No Plug; 230 VAC 5140204 Mfr # SI-B101
COMPARE
2400 230 Bead Genie™ Vortex Shaker, 250 to 2400 rpm, No Plug; 230 VAC
$3,247.35CAD / Each
Ohaus Bead Genie™ Vortex Shaker, 250 to 2400 rpm, European Plug; 230 VAC 5140206 Mfr # SI-B102
COMPARE
2400 230 Bead Genie™ Vortex Shaker, 250 to 2400 rpm, European Plug; 230 VAC
$3,247.35CAD / Each
Scientific Industries Bead Genie™ Vortex Shaker, 250 to 2400 rpm, UK Plug; 230 VAC 5140207 Mfr # SI-B103
COMPARE
2400 230 Bead Genie™ Vortex Shaker, 250 to 2400 rpm, UK Plug; 230 VAC
$3,247.35CAD / Each