Ohaus Adventure AX Series Semi-Micro Analytical Balances

Stability, accuracy, and fast operation ensure optimal weighing results
  • GLP/GMP compliance via real-time clock
  • Dual USB ports in front and rear offer easy access and connection
  • Color touch screen provides simple and fast operation with or without gloves
  • Six mechanical keys allow operator to perform repetitive tasks
  • AutoCal™ System offers automatic internal calibration
  • Extra wide draft shield doors provide access to larger weighing vessels
  • Overall design allows for easy cleaning, minimizing contamination
The Adventure AX-series semi-micro analytical balances are designed for high-precision weighing in laboratories, industrial, and educational environments. They feature various weighing modes, including part counting, percent weighing, dynamic, density, check weighing, totalization, and formulation.

These balances offer a user-friendly color touchscreen with an intuitive icon-based interface, allowing easy operation even with gloves. They also include three-level user management for compliance with GLP and GMP standards. Connectivity is enhanced with two USB ports for data transfer and backup, and options for both internal and external calibration, including an AutoCal System for automatic calibration.

The balances are durable, with a large weighing surface, a space-saving draft shield, and six mechanical keys for ease of use. Additional features include a cloning function for replicating settings, RS 232 interface, integral weigh below hook, and multiple customization options such as selectable environmental settings, audible indicators, and language preferences. The balances are built to ensure consistent performance and compliance in diverse settings.
$10,696.81 - $13,740.84CAD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Repeatability (mg)
Availability Pricing
Ohaus AX85 Adventurer Semi-Micro Balance, 82g x 0.01mg with Internal Calibration 1160129 Mfr # AX85
COMPARE
Internal 82 0.02
$10,696.81CAD / Each
Ohaus AX125D Adventurer Semi-Micro Balance, 82g/120g x 0.01mg/0.1mg with Internal Calibration 1160130 Mfr # AX125D
COMPARE
Internal 120 0.1
$11,415.83CAD / Each
Ohaus AX225D Adventurer Semi-Micro Balance, 102g/220g x 0.01mg/0.1mg with Internal Calibration 1160131 Mfr # AX225D
COMPARE
Internal 220 0.1
$13,740.84CAD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat