Multi-Channel Thermocouple Data Loggers with LCD
QuadTemp2000 and OctTemp2000
  • Large backlit LCD
  • Multi-channel recording
  • Front panel keypad
  • Real-time operation
  • On-screen statistics
  • High-speed download
MadgeTechs’ multi-channel thermocouple based display loggers are popular choices for monitoring temperature in multiple locations. The units accept J, K, T, E, R, S, B, and N thermocouples. All versions take synchronous samples for the highest data fidelity.
The QuadTemp2000 is a compact device that can measure and record almost 1 million readings per channel (all channels recorded). The QuadTemp2000 builds on the success of the original QuadTemp but improves the reading rate to 10Hz, 48X faster download speed (115,200 baud), 40X increased memory capacity, and selectable active channels for maximizing memory use.
The OctTemp2000 is an eight-channel thermocouple temperature logger with similar capabilities as the QuadTemp2000, but with 8 thermocouple channels. Similarly, the OctTemp2000 i
$1,821.11CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
MadgeTech OctTemp2000 Eight-Channel Thermocouple Recorder with LCD RK-23000-33 Mfr # 902172-00
COMPARE
$1,821.11CAD / Each