MadgeTech OctTemp2000 Eight-Channel Thermocouple Recorder with LCD
Representative image only.
MadgeTech OctTemp2000 Eight-Channel Thermocouple Recorder with LCD
MadgeTech | Mfr: 902172-00| Item: 2300033
QuadTemp2000 and OctTemp2000
$2,048.01CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$2,048.01CAD / Each
Buy It Together
Current Item
MadgeTech OctTemp2000 Eight-Channel Thermocouple Recorder with LCD
MadgeTech OctTemp2000 Eight-Channel Thermocouple Recorder with LCD
2300033
$2,048.01CAD / Each
+ + =
Total Price 2745.66 ADD 3 ITEMS TO CART
Product Description
Includes
Data logger comes with power adapter.
Key Features
  • Large backlit LCD
  • Multi-channel recording
  • Front panel keypad
  • Real-time operation
  • On-screen statistics
  • High-speed download
  • 1 year warranty
More About this Item
MadgeTechs’ multi-channel thermocouple based display loggers are popular choices for monitoring temperature in multiple locations. The units accept J, K, T, E, R, S, B, and N thermocouples. All versions take synchronous samples for the highest data fidelity.
The QuadTemp2000 is a compact device that can measure and record almost 1 million readings per channel (all channels recorded). The QuadTemp2000 builds on the success of the original QuadTemp but improves the reading rate to 10Hz, 48X faster download speed (115,200 baud), 40X increased memory capacity, and selectable active channels for maximizing memory use.
The OctTemp2000 is an eight-channel thermocouple temperature logger with similar capabilities as the QuadTemp2000, but with 8 thermocouple channels. Similarly, the OctTemp2000 i
1 Year Warranty
Specifications
  • Number of Channels8
  • Input TypeTemperature (J, K, T, E, R, S, B, or N thermocouple)
  • Temperature Accuracy (° C)±0.5 (0 to +50)
  • Max Thermocouple (° C)1820
  • Min Thermocouple (° C)-270
  • Thermocouple Resolution (° C)0.1
  • Memory Size256,000/channel
  • Length (in)7 1/4
  • Height (in)2 45/64
  • Width (in)1 11/16
  • DescriptionEight-Channel Thermocouple Recorder with LCD