Mini Temperature and Humidity Dataloggers
Provide consice measurements for critical environmental conditions
  • Includes easy-to-use user configurable software
  • Units accurate to ±1°F, (±0.5°C)
These mini dataloggers are ideal for recording temperature and humidity conditions in labs, food storage facilities, warehouses, refrigerators, freezers, museums, greenhouses and more.
Features include high/low alarms, PC, delayed start or pushbutton control, user selectable stop on memory full, or memory rollover for continuous recording. Included software allows you to view graph and text simultaneously and provides the ability to calibrate dataloggers.
Units provide feature a standard USB interface, and includes software that is Windows® 98SE, ME, 2000, XP and above compatible.
$222.85 - $302.41CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Input Type Max Temperature (° F) Temperature Accuracy (° F)
Availability Pricing
Supco SL300TH Mini Temperature and Humidity Data Logger 2303972 Mfr # SL300TH
COMPARE
Temperature and humidity 160 ±1
$302.41CAD / Each
Supco SL300T Mini Temperature Data Logger 2303974 Mfr # SL300T
COMPARE
Temperature 185 ±1
$222.85CAD / Each