Millipore® Syringe Filters
Designed for maximum sample recovery
  • Millipore Express PES (polyethersulfone) offers the fastest filtration. It is specifically designed for HTP and provides low protein binding sterile filtration
  • Durapore® PVDF (polyvinylidene fluoride) provides the lowest protein binding (99% recovery); ideal for filtration of high value biomolecules (e.g. monoclonal antibodies)
  • MF-Millipore MCE (mixed cellulose esters) is the most widely used membrane for sterilization of aqueous solutions and removal of trace protein contamination
  • Millipore LCR PTFE (polytetrafluoroethylene) is made of unsupported hydrophilic PTFE, compatible with all commonly used HPLC solvents. It can be used for filter aqueous fluids without prior wetting
  • Nylon is hydrophilic and compatible with a broad range of solvents
  • Fluoropore PTFE is a hydrophobic PTFE membrane bonded to a high-density polyethylene support; ideal for clarification of organic solvents. Sterile, individually packed, pyrogen free
  • HPLC sample preparation filters are nonsterile. The 4-mm filter units feature a stepped outlet. All products have been tested for UV-absorbing extractables. Certificate included
  • Ion chromatography filters are tested for low binding and low extractables—certificate is included. They are used for removing particulates from aqueous and mild organic solvents for cleaner IC spectra. Each unit is individually packaged to minimize the risk of extraneous ionic contamination
Millipore syringe filters are available in a variety of membranes and housing styles to meet your application needs. Ideal for sterile, ion chromatography, and HPLC sample preparation. All syringe filters have a female luer-lock inlet and a male luer slip connector.
$413.06 - $885.73CAD / Pkg of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
EMD Millipore SLFGX13NL Syringe Filters, Fluoropore PTFE, 13 mm dia, 0.20 µm, 100/pk 2995070 Mfr # SLFGX13NL
COMPARE
13 Fluoropore PTFE 0.2
$702.00CAD / Pkg of 100
EMD Millipore SLFH025NS Syringe Filters, Fluoropore PTFE, 25 mm dia, 0.45 µm, 50/pk 2995074 Mfr # SLFH025NS
COMPARE
25 Fluoropore PTFE 0.45
$885.73CAD / Pkg of 50
EMD Millipore SLAP02550 Syringe Filters, Glass Fiber Prefilter 50pk 8105300 Mfr # SLAP02550
COMPARE
25 Glass Microfiber (GMF) 0.22
$413.06CAD / Pkg of 50
EMD Millipore SLGV004SL General Purpose Syringe Filter, Durapore PVDF, 0.22um, 4mm dia, 100/pk 8105304 Mfr # SLGV004SL
COMPARE
4 Durapore PVDF 0.22
$770.07CAD / Pkg of 100
EMD Millipore SLSV025LS Syringe Filters, Durapore PVDF, 5um, 25 mm dia, 50/pk 8105308 Mfr # SLSV025LS
COMPARE
25 Durapore PVDF 5
$616.91CAD / Pkg of 50
EMD Millipore SLHV033NS HPLC Certified Syringe Filter, Durapore PVDF, 0.45um, 33mm dia, 50/pk 8105324 Mfr # SLHV033NS
COMPARE
33 Durapore PVDF 0.45
$495.06CAD / Pkg of 50
EMD Millipore SLLHC13NL Syringe Filters, PTFE, .45µm 100pk 8105352 Mfr # SLLHC13NL
COMPARE
13 Fluoropore PTFE 0.45
$712.46CAD / Pkg of 100
EMD Millipore SLLHC25NS Syringe Filters, PTFE, .45µm 50pk 8105354 Mfr # SLLHC25NS
COMPARE
25 Fluoropore PTFE 0.45
$570.13CAD / Pkg of 50
Showing 1- 8 of 8