EMD Millipore SLFH025NS Syringe Filters, Fluoropore PTFE, 25 mm dia, 0.45 µm, 50/pk

Representative image only.

EMD Millipore SLFH025NS Syringe Filters, Fluoropore PTFE, 25 mm dia, 0.45 µm, 50/pk

EMD Millipore | Mfr: SLFH025NS| Item: 2995074

Designed for maximum sample recovery

$885.73CAD / Pkg of 50

$885.73CAD / Pkg of 50
Product Description

Key Features

  • Millipore Express PES (polyethersulfone) offers the fastest filtration. It is specifically designed for HTP and provides low protein binding sterile filtration
  • Durapore® PVDF (polyvinylidene fluoride) provides the lowest protein binding (99% recovery); ideal for filtration of high value biomolecules (e.g. monoclonal antibodies)
  • MF-Millipore MCE (mixed cellulose esters) is the most widely used membrane for sterilization of aqueous solutions and removal of trace protein contamination
  • Millipore LCR PTFE (polytetrafluoroethylene) is made of unsupported hydrophilic PTFE, compatible with all commonly used HPLC solvents. It can be used for filter aqueous fluids without prior wetting
  • Nylon is hydrophilic and compatible with a broad range of solvents
  • Fluoropore PTFE is a hydrophobic PTFE membrane bonded to a high-density polyethylene support; ideal for clarification of organic solvents. Sterile, individually packed, pyrogen free
  • HPLC sample preparation filters are nonsterile. The 4-mm filter units feature a stepped outlet. All products have been tested for UV-absorbing extractables. Certificate included
  • Ion chromatography filters are tested for low binding and low extractables—certificate is included. They are used for removing particulates from aqueous and mild organic solvents for cleaner IC spectra. Each unit is individually packaged to minimize the risk of extraneous ionic contamination
  • Fluoropore PTFE (polytetrafluoroethylene) is a hydrophobic PTFE membrane bonded to a high-density polyethylene support; ideal for clarification of organic solvents. Sterile, individually packaged, pyrogen free
  • 1 year warranty

More About this Item

Millipore syringe filters are available in a variety of membranes and housing styles to meet your application needs. Ideal for sterile, ion chromatography, and HPLC sample preparation. All syringe filters have a female luer-lock inlet and a male luer slip connector.

HPLC sample preparation filters are nonsterile. The 4-mm units feature a stepped outlet. All products have been tested for UV–absorbing extractables. Certificate included.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year Warranty
Specifications
  • Membrane MaterialFluoropore PTFE
  • Pore Size (µm)0.45
  • Membrane Diameter (mm)25
  • Housing MaterialHigh-density Polyethylene (HDPE)
  • DescriptionSyringe Filters, Fluoropore PTFE, 25 mm dia, 0.45 µm, 50/pk
  • Warranty1 year
Reviews
Q&A