Mettler Toledo EVA Volumetric Karl Fischer Titrators

Efficiently determine the water content of simple liquids, soluble solids, and more challenging insoluble samples
  • Fast reaction rate without sacrificing accuracy
  • High precision—exceptional dispensing resolution of 120,000 steps
  • Modular system snaps in quickly—easily reconfigure burettes, sensors, and cells as workflows evolve
  • Easy-to-understand One Click™ touchscreen user interface
  • Save shortcuts to minimize errors and save time
  • Burette with RFID chip saves all relevant titrant data
  • Solvent manager provides safe chemical handling
  • Flexible data export via USB ports or Ethernet
Ensure the quality of your product by accurately determining the water content in your sample. The EVA Karl Fischer titrators simplify sample handling and eliminate the risk of human error, delivering highly reproducible and accurate results within a matter of minutes. Volumetric titrators analyze liquids, soluble solids, and insoluble solids (EVA V3 only) with water content between 100 ppm to 100% in single measurements.

A combination of the Fast Forecasting Amperometric (FFA™) control algorithm, a fast-response digital sensor, state-of-the-art dispensing technology, and low-drift glass titration vessels ensure fast and accurate results. A short stir bar and the conical shape of the vessel lead to high turbulence, effectively dispersing the titrant for a reliable measurement. Use the large color touchscreen to operate the simple-to-use interface to customize shortcuts and simply daily tasks.

Automatic sensor and burette recognition reduces extra data entry steps and helps minimize errors. Burettes feature an RFID chip that contains concentration or lifetime of the titrant and is directly transferred to the titrator. Ensure user safety with the solvent manager. It is fully controlled by the titrator, and offers filling, draining, and solvent exchange without additional user interaction. For secure titrator operation, designate user groups to allow full or limited access. Easily export data in CSV, PDF, or XML files using a memory stick, connect to a printer via the USB port, or network export via Ethernet.
2 PRODUCT OPTIONS
Compareitem specifications
Applications
Availability Pricing
Mettler Toledo 30869280 EVA V1 Volumetric Karl Fischer Titrator for Liquids and Soluble Solids 3333209 Mfr # 30869280
COMPARE
Liquids and soluble solids
$19,183.54CAD / Each
Mettler Toledo 30869282 EVA V3 Volumetric Karl Fischer Titrator for Liquids, Soluble Solids, and Insoluble Solids 3333210 Mfr # 30869282
COMPARE
Liquids, soluble solids, and insoluble solids
$26,462.08CAD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat