Mettler Toledo 30869280 EVA V1 Volumetric Karl Fischer Titrator for Liquids and Soluble Solids

Representative image only.

Mettler Toledo 30869280 EVA V1 Volumetric Karl Fischer Titrator for Liquids and Soluble Solids

Mettler Toledo | Mfr: 30869280| Item: 3333209

Efficiently determine the water content of simple liquids, soluble solids, and more challenging insoluble samples

$18,800.46CAD / Each

$18,800.46CAD / Each

Buy It Together

Current Item

Mettler Toledo 30869280 EVA V1 Volumetric Karl Fischer Titrator for Liquids and Soluble Solids
Mettler Toledo 30869280 EVA V1 Volumetric Karl Fischer Titrator for Liquids and Soluble Solids
3333209

$18,800.46CAD / Each
+ =
Total Price 21040.01 ADD 2 ITEMS TO CART
Product Description

Includes

5 mL burette set, titration kit, solvent pump dPump KF kit, burette drive dDrive with cable, dSens M143 sensor kit, titrator cover, touchscreen protective cover, terminal cable, dSens dVP4-T cable, external power supply, and power cable.

Key Features

  • Fast reaction rate without sacrificing accuracy
  • High precision—exceptional dispensing resolution of 120,000 steps
  • Modular system snaps in quickly—easily reconfigure burettes, sensors, and cells as workflows evolve
  • Easy-to-understand One Click™ touchscreen user interface
  • Save shortcuts to minimize errors and save time
  • Burette with RFID chip saves all relevant titrant data
  • Solvent manager provides safe chemical handling
  • Flexible data export via USB ports or Ethernet
  • Use for determining water content of liquid and soluble solid samples
  • 1 year warranty

More About this Item

Ensure the quality of your product by accurately determining the water content in your sample. The EVA Karl Fischer titrators simplify sample handling and eliminate the risk of human error, delivering highly reproducible and accurate results within a matter of minutes. Volumetric titrators analyze liquids, soluble solids, and insoluble solids (EVA V3 only) with water content between 100 ppm to 100% in single measurements.

A combination of the Fast Forecasting Amperometric (FFA™) control algorithm, a fast-response digital sensor, state-of-the-art dispensing technology, and low-drift glass titration vessels ensure fast and accurate results. A short stir bar and the conical shape of the vessel lead to high turbulence, effectively dispersing the titrant for a reliable measurement. Use the large color touchscreen to operate the simple-to-use interface to customize shortcuts and simply daily tasks.

Automatic sensor and burette recognition reduces extra data entry steps and helps minimize errors. Burettes feature an RFID chip that contains concentration or lifetime of the titrant and is directly transferred to the titrator. Ensure user safety with the solvent manager. It is fully controlled by the titrator, and offers filling, draining, and solvent exchange without additional user interaction. For secure titrator operation, designate user groups to allow full or limited access. Easily export data in CSV, PDF, or XML files using a memory stick, connect to a printer via the USB port, or network export via Ethernet.

Conformite Europeenne1 Year Warranty
Specifications
  • Units%, mg, mg/g, mg/kg, mg/mL, ppm, g/100g, g/100mL, g/kg, g/mL, µg/L, µg/mL, µg
  • ApplicationsLiquids and soluble solids
  • Display Type7" TFT IPS color display with capacitive multitouch screen
  • Display Resolution1024 x 600 pixels
  • Min Temperature (° C)18
  • Max Temperature (DegreeC)28
  • OutputUSB (2) and Ethernet
  • Data LoggingYes
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Length (in)7
  • Width (in)5 5/16
  • Height (in)7 1/4
  • Length (cm)17.7
  • Width (cm)13.5
  • Height (cm)18.5
  • DescriptionEVA V1 Volumetric Karl Fischer Titrator for Liquids and Soluble Solids
  • Warranty1 year
Reviews
Q&A