Mettler Toledo Density2Go Density Meters
Measure density anywhere—lab, plant floor, outdoors
  • Built-in sampling pump automates the filling and emptying of the sample cell
  • Bright color display and backlit sample cell let you take accurate measurements in any lighting conditions
  • Lightweight, ergonomic design accommodates single-handed operation, right or left hand compatible
  • Store 1100 measurements in internal memory
This digital, portable density meter makes taking measurements outside of a laboratory setting quick and easy. Whether you are measuring samples at production intervals or out in the field, you will be confident you have accurate results. Minimize errors and save time with the automatic internal sampling pump. Simply push up on the joystick and the pump fills the sample chamber. Brightly colored display with intuitive menu-driven interface and glove-friendly single-handed operation offer convenience and ease of use. Backlit sample cell lets you spot bubbles or other artifacts that may affect measurements. Rugged yet light and ergonomic construction allows frequent testing for extended periods of time without suffering from fatigue.
$5,915.66 - $6,586.05CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Density Range (g/cm³) Description
Availability Pricing
Mettler Toledo 30330857 Densito Handheld Density Meter 2575330 Mfr # 30330857
COMPARE
3 Densito Handheld Density Meter
$5,915.66CAD / Each
Mettler Toledo 30330858 DensitoPro Handheld Density Meter with RFID and Barcode Reader 2575331 Mfr # 30330858
COMPARE
3 DensitoPro Handheld Density Meter with RFID and Barcode Reader
$6,586.05CAD / Each