Mettler Toledo 30330858 DensitoPro Handheld Density Meter with RFID and Barcode Reader
Representative image only.
Mettler Toledo 30330858 DensitoPro Handheld Density Meter with RFID and Barcode Reader
Mettler Toledo | Mfr: 30330858| Item: 2575331
Measure density anywhere—lab, plant floor, outdoors
$6,586.05CAD / Each
$6,586.05CAD / Each
Buy It Together
Current Item
Mettler Toledo 30330858 DensitoPro Handheld Density Meter with RFID and Barcode Reader
Mettler Toledo 30330858 DensitoPro Handheld Density Meter with RFID and Barcode Reader
2575331
$6,586.05CAD / Each
+ =
Total Price 6725.87 ADD 2 ITEMS TO CART
Product Description
Includes
Filling tube assembly, USB-C cable, density standards, rechargeable Li-ion battery, and universal power supply with plug adapters. DensitoPro also includes ten SmartSample RFID tags.
Key Features
  • Built-in sampling pump automates the filling and emptying of the sample cell
  • Bright color display and backlit sample cell let you take accurate measurements in any lighting conditions
  • Lightweight, ergonomic design accommodates single-handed operation, right or left hand compatible
  • Store 1100 measurements in internal memory
  • RFID and barcode reader provides point-and-shoot sample identification (DensitoPro only)
  • 1 year warranty
More About this Item
This digital, portable density meter makes taking measurements outside of a laboratory setting quick and easy. Whether you are measuring samples at production intervals or out in the field, you will be confident you have accurate results. Minimize errors and save time with the automatic internal sampling pump. Simply push up on the joystick and the pump fills the sample chamber. Brightly colored display with intuitive menu-driven interface and glove-friendly single-handed operation offer convenience and ease of use. Backlit sample cell lets you spot bubbles or other artifacts that may affect measurements. Rugged yet light and ergonomic construction allows frequent testing for extended periods of time without suffering from fatigue.
Sample identification using the DensitoPro density meter's RFID and barcode reader ensures data integrity for reporting. RFID reading function is compatible with Mettler Toledo's SmartSample tags (included with DensitoPro). Barcode reading function recognizes linear barcodes only.
1 Year WarrantyConformite Europeenne
Specifications
  • Min Density Range (g/cm³)0
  • Max Density Range (g/cm³)3
  • Density Resolution (g/cm³)0.001
  • Density Accuracy±0.001 g/cm³
  • Max Temperature (° F)122
  • Min Temperature (° F)32
  • Max Temperature (° C)50
  • Min Temperature (° C)0
  • Internal Memory1100
  • Power (VAC)100 to 260
  • Battery Type1 rechargeable lithium (included)
  • DescriptionDensitoPro Handheld Density Meter with RFID and Barcode Reader