Megger Test Leads for Resistance Meters

Select the test lead set(s) for your application.
$589.36 - $2,818.60CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Megger 242021-7 Potential Test Lead, Single Hand Spike, 7 ft 2001103 Mfr # 242021-7
COMPARE
Potential Test Lead, Single Hand Spike, 7 ft
$1,166.34CAD / Each
Megger 242011-18 LEADS HELICAL SPIKES 10A 18FT 2001105 Mfr # 242011-18
COMPARE
$2,818.60CAD / Each
Megger 242002-18 LEADS; HAND SPIKES 10A 18FT 2001109 Mfr # 242002-18
COMPARE
$2,192.40CAD / Each
Megger 242005-7 Duplex Test Leads with 0.5" Kelvin Clips, 10 A, 7 ft 2001114 Mfr # 242005-7
COMPARE
Duplex Test Leads with 0.5" Kelvin Clips, 10 A, 7 ft
$589.36CAD / Each
Megger 242006-7 Duplex Test Leads with 1.5" Kelvin Clips, 10 A, 7 ft 2001115 Mfr # 242006-7
COMPARE
Duplex Test Leads with 1.5" Kelvin Clips, 10 A, 7 ft
$868.86CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat