Mark-10 Testing Modules for IntelliMESUR® Software
Enhance your digital test frame with IntelliMESUR software with advanced feature modules
  • Field upgradeable—no need to ship it to the factory or call for service
Characterize and analyze material behavior and measure static and dynamic coefficient of friction (COF) with optional software upgrades. Upgrades can be done at any time and easily upgraded in the field via activation code.
$825.10 - $4,508.57CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Mark-10 Coefficient of Friction (COF) Testing Module for IntelliMESUR® Software 6780108 Mfr # IMF001
COMPARE
Test frames with IntelliMESUR® software Coefficient of Friction (COF) Testing Module
$825.10CAD / Each
Mark-10 Materials Testing Calculations Module for IntelliMESUR® Software 6780109 Mfr # IMF002
COMPARE
Test frames with IntelliMESUR® software Materials Testing Calculations Module for IntelliMESUR® Software
$4,508.57CAD / Each