Mark-10 Materials Testing Calculations Module for IntelliMESUR® Software
Representative image only.
Mark-10 Materials Testing Calculations Module for IntelliMESUR® Software
Mark-10 | Mfr: IMF002| Item: 6780109
Enhance your digital test frame with IntelliMESUR software with advanced feature modules
$4,508.57CAD / Each
$4,508.57CAD / Each
Product Description
Key Features
  • Field upgradeable—no need to ship it to the factory or call for service
  • Characterize and analyze the behavior of materials, components, and assemblies
More About this Item
Characterize and analyze material behavior and measure static and dynamic coefficient of friction (COF) with optional software upgrades. Upgrades can be done at any time and easily upgraded in the field via activation code.
The materials testing module features a suite of common calculations such as tensile strength, Young's modulus, yield point, offset yield strength, ultimate strength, and rupture.
Specifications
  • Compatible WithTest frames with IntelliMESUR® software
  • DescriptionMaterials Testing Calculations Module for IntelliMESUR® Software
  • Warranty3 years