Mark-10 Series F Digital Force Test Frames with EasyMESUR® Control Panel
Easily set up and control tests using the EasyMESUR integrated touchscreen control panel
  • Virtually limitless applications—use for peel testing, wire tensile testing, compression spring testing, and much more!
  • Works with Series F force sensors to deliver ±0.2% of full-scale accuracy
  • Directly mount sensors to frames—no connecting cables required
  • Fast data collection—collects load and travel data at up to 100 Hz
  • Two USB ports for saving, exporting, and recalling of data
  • Rigid and precise mechanical design
  • Solid-state upper and lower limit switches
  • Password protection limits access to test editing and calibration
  • Field upgradeable—easily add optional functionalities via activation codes
Use these force test frames to test the tensile strength and compression force in various industries, including food packaging, electronics, and aerospace manufacturing. The Series F digital force test frames work in conjunction with Series F force sensors for force testing applications up to 2.2 kN (500 lbF). The sensors mount directly on the crosshead of the frame without any connecting cables.
Digital force test frames feature the EasyMESUR control panel to easily set up tests, view results, and export data. Use the 7" touchscreen to create and run test to a load, test to a distance, and average load testing. Select to view a test run in standard view, graph view, or results view depending on your application. Use the solid-state upper and lower limit switches graduated in inch and mm scale as test limits or as test exceptions.
Easily add more advanced functionalities including cycling, break detection, distance measurement, graphing, reporting, and much more via activation codes. EasyMESUR control panel saves up to 100,000 data points per run. Save individual run data or results files to a USB drive or internal storage accessible from a PC. Export data and results as .csv files (graphing & reporting option required)—order Complete Options Package 67801-17 that includes the graphing and reporting functions. Rigid extrusion-based frames provide exceptional stiffness, ensuring accuracy; test frames and force sensors offer position accuracy of ±0.15 mm (±0.006") under any load, at any position.
$5,525.21 - $8,192.05CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kN) Capacity (lbs) Description
Availability Pricing
Mark-10 Test Frame with EasyMESUR® Control Panel, Vertical, 0.5 kN / 100 lbF 6780110 Mfr # F105-EM
COMPARE
0.5 100 Test Frame with EasyMESUR® Control Panel, Vertical, 0.5 kN / 100 lbF
$5,525.21CAD / Each
Mark-10 Test Frame with EasyMESUR® Control Panel, Vertical, 1.3 kN / 300 lbF 6780111 Mfr # F305-EM
COMPARE
1.3 300 Test Frame with EasyMESUR® Control Panel, Vertical, 1.3 kN / 300 lbF
$5,819.89CAD / Each
Mark-10 Test Frame with EasyMESUR® Control Panel, Vertical, 2.2 kN / 500 lbF 6780112 Mfr # F505-EM
COMPARE
2.2 500 Test Frame with EasyMESUR® Control Panel, Vertical, 2.2 kN / 500 lbF
$7,278.55CAD / Each
Mark-10 Test Frame with EasyMESUR® Control Panel, Horizontal, 2.2 kN / 500 lbF 6780113 Mfr # F505H-EM
COMPARE
2.2 500 Test Frame with EasyMESUR® Control Panel, Horizontal, 2.2 kN / 500 lbF
$8,192.05CAD / Each