MAGic Clamp™ Incu-Shaker and Incu-Shaker Mini Accessories
Optimize performance of your shaking incubator
Change the capacitiy and capabilities of your shaker with these parts and accessories. Compatible with Benchmark Incu-Shaker™ Mini Incubating Shakers sold separately as 51705-01 and 51705-03.
The convenient MAGic Clamp™ platform is suitable for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
$64.09 - $355.82CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Description
Availability Pricing
MAGic Clamp H1000-MR-500 Magnetic Clamp, Erlenmeyer 500 ml. 5170516 Mfr # H1000-MR-500
COMPARE
500 mL Magnetic Clamp, Erlenmeyer Flask; 500 mL
$78.97CAD / Each
MAGic Clamp H1000-MR-250 Magnetic Clamp, Erlenmeyer 250 ml. 5170517 Mfr # H1000-MR-250
COMPARE
250 mL Magnetic Clamp, Erlenmeyer Flask; 250 mL
$78.97CAD / Each
MAGic Clamp H1000-MR-125 Magnetic Clamp, Erlenmeyer 125 ml. 5170518 Mfr # H1000-MR-125
COMPARE
125 mL Magnetic Clamp, Erlenmeyer Flask; 125 mL
$70.28CAD / Each
MAGic Clamp H1000-MR-50 Magnetic Clamp, Erlenmeyer 50 ml. 5170519 Mfr # H1000-MR-50
COMPARE
50 mL Magnetic Clamp, Erlenmeyer Flask; 50 mL
$64.09CAD / Each
MAGic Clamp H1000-MR-3000 Magnetic Clamp, Erlenmeyer 3000 ml. 5170521 Mfr # H1000-MR-3000
COMPARE
3000 mL Magnetic Clamp, Erlenmeyer Flask; 3000 mL
$238.32CAD / Each
MAGic Clamp H1000-MR-2000 Magnetic Clamp, Erlenmeyer 2000 ml. 5170523 Mfr # H1000-MR-2000
COMPARE
2000 mL Magnetic Clamp, Erlenmeyer Flask; 2000 mL
$151.76CAD / Each
MAGic Clamp H1000-MR-1000 Magnetic Clamp, Erlenmeyer 1000 ml. 5170526 Mfr # H1000-MR-1000
COMPARE
1000 mL Magnetic Clamp, Erlenmeyer Flask; 1000 mL
$119.20CAD / Each
MAGic Clamp H1000-MR-MP Microplate Magnetic Clamp, Holds 1. 5170527 Mfr # H1000-MR-MP
COMPARE
1 Microplate Microplate Magnetic Clamp
$167.23CAD / Each
MAGic Clamp H1000-MR-1550 Tube Rack, Holds 30 x 15 ml and 20 x 50 ml Tubes. 5170528 Mfr # H1000-MR-1550
COMPARE
30 x 15 mL and 20 x 50 mL Tube Rack; Holds 30 x 15 mL and 20 x 50 mL Tubes
$306.47CAD / Each
MAGic Clamp H1000-MR-T15 Tube Rack, Holds 32 x 15 ml Tubes. 5170529 Mfr # H1000-MR-T15
COMPARE
32 x 15 mL Tube Rack; Holds 32 x 15 mL Tubes
$306.47CAD / Each
MAGic Clamp H1000-MR-T50 Tube Rack, Holds 15 x 50 ml Tubes. 5170531 Mfr # H1000-MR-T50
COMPARE
15 x 50 mL Tube Rack; Holds 15 x 50 mL Tubes
$306.47CAD / Each
MAGic Clamp H1000-MR-T600 Tube Rack, Holds 4 x 500 ml or 600 ml Conical Bottles. 5170532 Mfr # H1000-MR-T600
COMPARE
500/600 mL Tube Rack; Holds 4 x 500/600 mL Conical Bottles
$355.82CAD / Each
MAGic Clamp H1000-MR-T150H Horizontal Tube Rack, Holds 4 x 15 ml Tubes. 5170534 Mfr # H1000-MR-T150H
COMPARE
4 x 15 mL Horizontal Tube Rack; Holds 4 x 15 mL Tubes
$216.59CAD / Each
MAGic Clamp H1000-MR-T500H Horizontal Tube Rack, Holds 2 x 50 ml Tubes. 5170536 Mfr # H1000-MR-T500H
COMPARE
2 x 50 mL Horizontal Tube Rack; Holds 2 x 50 mL Tubes
$216.59CAD / Each
Benchmark Scientific H1000-MR-CMB Combo for 125 mL, 250 mL, and 500 mL 5170537 Mfr # H1000-MR-CMB
COMPARE
125-, 250-, or 500-mL Adjustable Flask Clamp, for 125-, 250-, or 500-mL Flasks; 1/Ea
$83.54CAD / Each
Showing 1- 15 of 15