MadgeTech RFTEMP2000A Series Wireless Temperature Data Loggers
Two-way wireless communication eliminates need to retrieve logger for maintenance
  • Data can stream live to a PC or transmit in batch form to accommodate any application
  • Programmable triggers collect data only when temperature exceeds the setpoint to preserve memory
  • Four-line LCD shows current reading and statistics for spot checks
Required UHF transceiver (18003-68) communicates with logger up to 500-ft away indoors, allowing PC control from across the room or from another room. Alarms for temperature include audible and visible alerts from the data logger, and alarm messages via the software in email or SMS text form. Wireless data loggers can be used as standalone items or in concert with up to 63 other loggers per transceiver for an integrated system. Use the software included with the transceiver to program and calibrate the logger, download data, analyze with statistics, and export to Microsoft Excel®.
$545.89 - $909.08CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Input Type Max Temperature (° F) Temperature Resolution (° F)
Availability Pricing
MadgeTech RFTemp2000A Wireless Temperature Data Logger 1801300 Mfr # RFTEMP2000A
COMPARE
Temperature (J, K, T, E, R, S, B, or N thermocouple) 140 0.018
$545.89CAD / Each
MadgeTech RFTCTemp2000A Wireless Datalogger for Thermocouples, mini connector 1801301 Mfr # RFTCTEMP2000A-MP
COMPARE
Temperature (J, K, T, E, R, S, B, or N thermocouple) 140 0.18
$581.25CAD / Each
MadgeTech RFRTDTemp2000A Wireless Data Logger for RTDs 1801303 Mfr # RFRTDTEMP2000A
COMPARE
Temperature (RTD) 1562 0.018
$585.67CAD / Each
MadgeTech RFPRHTemp2000A Wireless Data Logger, temperature, humidity, pressure 1801304 Mfr # RFPRHTEMP2000A
COMPARE
Temperature, humidity, and pressure 140 0.018
$909.08CAD / Each