MadgeTech Extended-Life Temperature/Humidity/Pressure Data Loggers
10-year battery life for long-term, maintenance-free operation in the field
  • Real-time recording with 1-second reading rate
  • Programmable high and low limits; alarm is activated when parameter reaches or exceeds set limits
  • Red/green LEDs indicate logging, start mode, low battery, full memory, and alarm condition
  • Ultrahigh-speed download capability
  • Software package (required) provides easy setup, operation, and data download for later analysis
  • Multiple start/stop mode lets you start and stop the device multiple times without having to download data or communicate with a PC
  • ABS plastic case handles temperatures from –40 to 176°F (–40 to 80°C)
  • Optional memory wrap function and password protection
Monitors pressure, humidity, and temperature levels in clean rooms, warehouses, storage rooms, and restoration projects.
$221.38 - $563.57CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
MadgeTech Temp101A Temperature Extended-Life Data Logger 7000101 Mfr # TEMP101A
COMPARE
175 ±0.9 0.018
$221.38CAD / Each
MadgeTech RHTemp101A Temperature and Relative Humidity Extended-Life Data Logger 7000102 Mfr # RHTEMP101A
COMPARE
175 ±0.9 0.018
$351.40CAD / Each
MadgeTech TC101A-MP Thermocouple Temperature Extended-Life Data Logger 7000103 Mfr # TC101A-MP
COMPARE
±0.45 0.18
$369.82CAD / Each
MadgeTech PRHTEMP101A Pressure, Humidity & Temperature Extended-Life Data Logger 7000244 Mfr # PRHTEMP101A
COMPARE
175 ±0.9 0.018
$563.57CAD / Each