Low Form Glass Dewar Flasks for Liquid Nitrogen
These Dewars permit operations not practical with standard dewars
  • Small flasks can be inserted into Dewars
  • Rapid freezing and recovery of tissue/organ specimens
  • Agitation can be provided with a magnetic stirrer
These Dewar flasks expand the applications of standard cylindrical or spherical Dewars. The protective aluminum base on both sides extends the full height of the Dewars. Protected vacuum tip-off tubulations are off to the side to minimize space between the bottoms and magnetic stirrers.
$632.08 - $1,863.84CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Volume (mL)
Availability Pricing
COMPARE
275
$632.08CAD / Each
COMPARE
600
$732.27CAD / Each
COMPARE
1000
$785.32CAD / Each
COMPARE
2000
$1,863.84CAD / Each