Lovibond EComparator Series Colorimeters
Advance from visual to electronic color measurement with confidence
  • Interpret results easily with visual color comparison mode and direct digital reading
  • Obtain quality checks with the on-screen warning system—results displayed in green, yellow, and red
  • Easily set time, date, preferences, and projects through the intuitive touch screen
  • Save 20,000 individual readings to the internal memory
  • Internationally recognized industry standards
These color meters cover a wide range of applications. The flexible pathlength lets you use the cuvettes that come with the meter, or up to a 50 mm cuvette. Additionally, the light shield helps ensure accurate readings by blocking ambient light. Units may be powered with four AA batteries or through the USB cable provided.
$7,381.68 - $13,503.62CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lovibond EComparator 2000 Series Colorimeter; Pt/Co 9957555 Mfr # 162010
COMPARE
Colorimeter; Pt/Co
$8,243.62CAD / Each
Lovibond EComparator 2000 Series Colorimeter; Gardner 9957560 Mfr # 162020
COMPARE
Colorimeter; Gardner
$7,381.68CAD / Each
Lovibond EComparator 3000 Series Colorimeter; ASTM 9957565 Mfr # 163010
COMPARE
Colorimeter; ASTM
$12,663.79CAD / Each
Lovibond EComparator 3000 Series Colorimeter; Saybolt 9957570 Mfr # 163020
COMPARE
Colorimeter; Saybolt
$13,503.62CAD / Each