Lovibond EComparator 2000 Series Colorimeter; Gardner
Representative image only.
Lovibond EComparator 2000 Series Colorimeter; Gardner
Lovibond | Mfr: 162020| Item: 9957560
Advance from visual to electronic color measurement with confidence
$7,381.68CAD / Each
$7,381.68CAD / Each
Buy It Together
Current Item
Lovibond EComparator 2000 Series Colorimeter; Gardner
Lovibond EComparator 2000 Series Colorimeter; Gardner
9957560
$7,381.68CAD / Each
+
Lovibond MD 600  Multiparameter Colorimeter 214020
9959900
$3,208.60CAD / Each
=
Total Price 10590.28 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
plastic and glass cuvettes, color reference standards, cell divider, screwdriver, USB power supply and cable, four AA batteries, and hard-sided carrying case.
Key Features
  • Interpret results easily with visual color comparison mode and direct digital reading
  • Obtain quality checks with the on-screen warning system—results displayed in green, yellow, and red
  • Easily set time, date, preferences, and projects through the intuitive touch screen
  • Save 20,000 individual readings to the internal memory
  • Internationally recognized industry standards
  • 1 year warranty
More About this Item
These color meters cover a wide range of applications. The flexible pathlength lets you use the cuvettes that come with the meter, or up to a 50 mm cuvette. Additionally, the light shield helps ensure accurate readings by blocking ambient light. Units may be powered with four AA batteries or through the USB cable provided.
1 Year Warranty
Specifications
  • DescriptionColorimeter; Gardner