Lightnin Small Batch, Direct-Drive Industrial Mixers
  • Mix materials with viscosities up to 10,000 cps
Use these versatile mixers for lab mixing or for scaling up pilot plant experiments. Use the fully adjustable tank-clamp to position mixer at any angle.
Use the selection guide (available in the Technical Library) to select the appropriate mixer for your application.
Includes fully adjustable tank-clamp to position mixer at any angle.
$4,825.35 - $6,652.36CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Shaft Length (in)
Availability Pricing
Lightnin 800013 Small Batch Direct-Drive Mixer, 1750 rpm, 1/20 HP XP Motor, 115V 0451820 Mfr # 800013
COMPARE
1750 29 24
$6,652.36CAD / Each
Lightnin 800000 Small batch mixer with two-speed motor, 115 VAC 0451830 Mfr # 800000
COMPARE
1600 42 24
$5,495.74CAD / Each
Lightnin 800129-1 Small batch fixed-speed mixer with 1/8-hp motor, 115 VAC 0451840 Mfr # 800129-1
COMPARE
1750 72 30
$4,825.35CAD / Each