Lightnin Heavy-Duty, Gear-Drive Industrial Batch Mixers
Ideal for mixing your high-viscosity materials
  • Maintains speeds from 20 to 350 rpm
  • Able to mix highly viscous fluids
These variable-speed, gear-drive mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
Gear-drive mixers mount with 20°> offset tank clamp (included). Available as either an air-powered or electric mixer.
Select from three speed options; 20 to 350 rpm (20:1 ratio), 20 to 280 rpm (14:1 ratio), or fixed speed of 350 or 280 rpm. The 3⁄4-hp mixers generate plenty of torque for your high-viscosity applications.
$5,038.99 - $10,203.23CAD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Motor Type
Availability Pricing
Lightnin EV5P25M1E36 Heavy-Duty Gear Drive Mixer, TENV, 20-350rpm, 720in-oz, 3/4" x 36" shaft 5031000 Mfr # EV5P25M1E36
COMPARE
350 720 TENV
$6,969.13CAD / Each
Lightnin EV5P25A1D36 Heavy-Duty Gear Drive Mixer, air, 20-350rpm, 720in-oz, 3/4" x 36" shaft 5031002 Mfr # EV5P25A1D36
COMPARE
350 720 Air-driven
$5,120.03CAD / Each
Lightnin EV5P50M2L36 Heavy-Duty Gear Drive Mixer, TENV, 20-350rpm, 1440in-oz, 3/4" x 36" shaft 5031004 Mfr # EV5P50M2L36
COMPARE
350 1440 TENV
$9,945.38CAD / Each
Lightnin EV5P50A2L36 Heavy-Duty Gear Drive Mixer, air, 20-350rpm, 1440in-oz, 3/4" x 36" shaft 5031006 Mfr # EV5P50A2L36
COMPARE
350 1440 Air-driven
$7,440.62CAD / Each
Lightnin EV5P25M1F48 Heavy-Duty Gear Drive Mixer, TENV, 20-350rpm, 720in-oz, 3/4" x 48" shaft 5031008 Mfr # EV5P25M1F48
COMPARE
350 720 TENV
$7,263.81CAD / Each
Lightnin EV5P25A1F48 Heavy-Duty Gear Drive Mixer, air, 20-350rpm, 720in-oz, 3/4" x 48" shaft 5031010 Mfr # EV5P25A1F48
COMPARE
350 720 Air-driven
$5,370.51CAD / Each
Lightnin EV6P50M2T48 Heavy-Duty Gear Drive Mixer, TENV, 20-280rpm, 1801in-oz, 3/4" x 48" shaft 5031012 Mfr # EV6P50M2T48
COMPARE
280 1801 TENV
$9,437.06CAD / Each
Lightnin EV6P50M2X60 Heavy-Duty Gear Drive Mixer, TENV, 20-280rpm, 1801in-oz, 3/4" x 60" shaft 5031016 Mfr # EV6P50M2X60
COMPARE
280 1801 TENV
$10,203.23CAD / Each
Lightnin EV5P25M1J60 Heavy-Duty Gear Drive Mixer, TENV, 20-350rpm, 3/4" x 60", 11.2" impeller 5031020 Mfr # EV5P25M1J60
COMPARE
350 720 TENV
$7,278.55CAD / Each
Lightnin EV5P25A1J60 Heavy-Duty Gear Drive Mixer, air, 20-350rpm, 720in-oz, 3/4" x 60" shaft 5031022 Mfr # EV5P25A1J60
COMPARE
350 720 Air-driven
$5,392.61CAD / Each
Lightnin EV6P75M2X60 Heavy-Duty Gear Drive Mixer, TENV, 20-280rpm, 2701in-oz, 3/4" x 60" shaft 5031028 Mfr # EV6P75M2X60
COMPARE
280 2701 TENV
$10,144.29CAD / Each
Lightnin EV5P50T2L36 Heavy-Duty Gear Drive Mixer, TEFC, 350rpm, 1440in-oz, 3/4" x 36" shaft 5031042 Mfr # EV5P50T2L36
COMPARE
350 1440 TEFC
$7,440.62CAD / Each
Lightnin EV5P25ET1F48 Heavy-duty, gear-drive mixer, XPRF 5031046 Mfr # EV5P25ET1F48
COMPARE
350 720 XPRF
$6,018.80CAD / Each
Lightnin EV5P25T1F48 Heavy-Duty Gear Drive Mixer, TEFC, 350rpm, 720in-oz, 3/4" x 48" shaft 5031048 Mfr # EV5P25T1F48
COMPARE
350 720 TEFC
$5,038.99CAD / Each
Lightnin EV5P25T1J60 Heavy-Duty Gear Drive Mixer, TEFC, 350rpm, 720in-oz, 3/4" x 60" shaft 5031054 Mfr # EV5P25T1J60
COMPARE
350 720 TEFC
$5,075.83CAD / Each
Lightnin EV6P75T2X60 Heavy-Duty Gear Drive Mixer, TEFC, 280rpm, 2701in-oz, 3/4" x 60" shaft 5031066 Mfr # EV6P75T2X60
COMPARE
280 2701 TEFC
$9,643.34CAD / Each
Lightnin EV5P25MIU60 Heavy-Duty Gear Drive Mixer, TENV, 20-350rpm, 3/4" x 60", 10" impeller 5032330 Mfr # EV5P25MIU60
COMPARE
350 720 TENV
$7,138.57CAD / Each
Lightnin EV5P5OAIP60 Heavy-Duty Gear Drive Mixer, air, 20-350rpm, 1440in-oz, 3/4" x 60" shaft 5032340 Mfr # EV5P5OAIP60
COMPARE
350 1440 Air-driven
$6,799.69CAD / Each
Showing 1- 18 of 18