LaMotte Single Test Colorimeter Kits
Single test, direct-reading colorimeters feature microprocessor technology
  • Analyzer features automatic wavelength selection for quick testing
Auto-zeroing and a hinged light cover make these meters easy to use. Ergonomic design fits in your hand comfortably. Water-resistant design protects your meter from damage. Download data to a computer or printer via the optional RS-232 interface.
$1,214.44 - $1,464.92CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
LaMotte DC-1500 Single Test Colorimeter Kit , Chlorine Free and Total " Liquid form 9956212 Mfr # RBY 3240
COMPARE
Single Test Colorimeter Kit , Chlorine Free and Total " Liquid form
$1,214.44CAD / Each
LaMotte DC-1500 Single Test Colorimeter Kit, Copper 9956218 Mfr # RBY 3245
COMPARE
Single Test Colorimeter Kit , Copper
$1,464.92CAD / Each