LaMotte DC-1500 Single Test Colorimeter Kit, Copper

Representative image only.

LaMotte DC-1500 Single Test Colorimeter Kit, Copper

LaMotte | Mfr: RBY 3245| Item: 9956218

Single test, direct-reading colorimeters feature microprocessor technology

$1,464.92CAD / Each

$1,464.92CAD / Each

Buy It Together

Current Item

LaMotte DC-1500 Single Test Colorimeter Kit, Copper
LaMotte DC-1500 Single Test Colorimeter Kit, Copper
9956218

$1,464.92CAD / Each
+ +
Environmental Express Model 200 Chlorine Colorimeter
9956178

$1,280.38CAD / Each
=
Total Price 5341.27 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

analyzer, glass sample vials, plastic water bottle, tablet crusher, test-specific reagent, foam-lined carrying case, and one 9 V battery.

Key Features

  • Analyzer features automatic wavelength selection for quick testing
  • 1 year warranty

More About this Item

Auto-zeroing and a hinged light cover make these meters easy to use. Ergonomic design fits in your hand comfortably. Water-resistant design protects your meter from damage. Download data to a computer or printer via the optional RS-232 interface.

1 Year Warranty
Specifications
  • Display TypeLCD
  • Light SourceLED
  • Detector TypeSilicon Photodiode
  • Test ParametersCopper
  • Number of Tests Included100
  • Data LoggingYes
  • Battery1 9V (included)
  • Length (in)6 1/2
  • Width (in)3 1/4
  • Height (in)2 1/2
  • DescriptionSingle Test Colorimeter Kit , Copper
  • Warranty1 year
Reviews
Q&A