Labnet BioPette Plus Autoclavable, Adjustable Volume Pipettes, Multi-Channel
Autoclavable, comfortable and accurate
  • Superior accuracy and reproducibility
  • Thumbwheel volume selection
  • Universal tip fit
The line of BioPette™ Plus pipettes are ergonomically designed for comfort and accuracy. Pipettes are engineered lightweight and well balanced, to be used in either the right or left hand. Large digital display provides enhanced visiblilty to the user reducing increasing accuracy. Chemically resistant, maintenance-free seals and finely polished pistons provide unsurpassed accuracy sample after sample.
Adjustable, digital micrometer is slightly recessed to prevent unintentional change and is protected from the effects of thermal expansion by the insulated body of the pipette. For convenience, volume can also be adjusted by turning the push-button. Ergonomically positioned, the tip ejector reduces fatigue during prolonged use. Pipettes have a universal shaft that will accept most pipette tips. The tip ejector is adjustable to accommodate various styles of tips and removable for pipetting into narrow tubes. Stainless steel tip ejector will not rust, corrode, or break. autoclavable.
$1,379.09 - $1,865.31CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
Labnet P4808-10 Pipette, 8-channel, autoclavable, 1-10 uL 2460116 Mfr # P4808-10
COMPARE
10 1 8
$1,418.87CAD / Each
Labnet P4508-50 Pipette, 8-channel, autoclavable, 5-50 uL 2460118 Mfr # P4808-50
COMPARE
50 5 8
$1,457.18CAD / Each
Labnet P4508-200 Pipette, 8-channel, autoclavable, 20-200 uL 2460120 Mfr # P4808-200
COMPARE
200 20 8
$1,865.31CAD / Each
Labnet P4508-300 Pipette, 8-channel, autoclavable, 50-300 uL 2460122 Mfr # P4808-300
COMPARE
300 50 8
$1,379.09CAD / Each
Labnet P4512-10 Pipette, 12-channel, autoclavable, 1-10 uL 2460124 Mfr # P4812-10
COMPARE
10 1 12
$1,700.29CAD / Each
Labnet P4512-200 Pipette, 12-channel, autoclavable, 20-200 uL 2460128 Mfr # P4812-200
COMPARE
200 20 12
$1,747.44CAD / Each
Labnet P4512-300 Pipette, 12-channel, autoclavable, 50-300 uL 2460130 Mfr # P4812-300
COMPARE
300 50 12
$1,697.35CAD / Each
Showing 1- 7 of 7