Labconco RXPert Class I Filtered Biological Safety Cabinets
Keep hazardous powders and particulates contained
  • Meet USP <800> requirements for nonsterile hazardous compounding with redundant filtration optional
  • Bag in/bag out HEPA filter provides protection during filter changes
The RXPert cabinet features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement. The filtered balance system is a Class I enclosure that complies with USP requirements for nonsterile compounding.
All models include a built-in analog airflow monitor. Some models include a stainless steel or epoxy work surface; a work surface is required if one is not included.
Single-Filtered Enclosures include an all-in-one factory-installed canopy and the damper allows for thimble ducting to the outside. All models require a remote blower, ductwork, and supporting base (order separately).
Compliance: USP <800>, SEFA 1-2010, modified ASHRAE 110-1995, ANSI Z9.5-2011, UL 61010-1, and CAN/CSA C22.2 No. 61010.1 standards.
$15,514.80 - $41,203.35CAD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exhaust Volume (CFM) Exterior Width (in) Description
Availability Pricing
Labconco 3971201 Filtered Balance System, Class I Biosafety Cabinet, SS Surface, 2' 3252134 Mfr # 3971201
COMPARE
145 24 Filtered Balance System, Class I Biosafety Cabinet, SS Surface, 2'
$16,295.69CAD / Each
Labconco 3971202 Filtered Balance System, Class I Biosafety Cabinet, Epoxy Surface, 2' 3252136 Mfr # 3971202
COMPARE
145 24 Filtered Balance System, Class I Biosafety Cabinet, Epoxy Surface, 2'
$15,514.80CAD / Each
Labconco 3971301 Filtered Balance System, Class I Biosafety Cabinet, SS Surface, 3' 3252140 Mfr # 3971301
COMPARE
220 36 Filtered Balance System, Class I Biosafety Cabinet, SS Surface, 3'
$18,225.83CAD / Each
Labconco 3971302 Filtered Balance System, Class I Biosafety Cabinet, Epoxy Surface, 3' 3252142 Mfr # 3971302
COMPARE
220 36 Filtered Balance System, Class I Biosafety Cabinet, Epoxy Surface, 3'
$17,142.89CAD / Each
Labconco 3971401 Filtered Balance System, Class I Biosafety Cabinet, SS Surface, 4' 3252146 Mfr # 3971401
COMPARE
295 48 Filtered Balance System, Class I Biosafety Cabinet, SS Surface, 4'
$20,273.85CAD / Each
Labconco 3971402 Filtered Balance System, Class I Biosafety Cabinet; Epoxy Surface, 4' 3252148 Mfr # 3971402
COMPARE
295 48 Filtered Balance System, Class I Biosafety Cabinet; Epoxy Surface, 4'
$18,866.76CAD / Each
Labconco RXPert  Tall Single Filtered Balance System, Stainless Steel Surface, 4', 115 V 3252154 Mfr # 3971476
COMPARE
380 48 Tall Single Filtered Balance System, Stainless Steel Surface, 4', 115 V
$22,579.70CAD / Each
Labconco RXPert  Tall Single Filtered Balance System, Epoxy Surface, 4', 115 V 3252156 Mfr # 3971477
COMPARE
380 48 Tall Single Filtered Balance System, Epoxy Surface, 4', 115 V
$21,165.25CAD / Each
Labconco RXPert  Tall & Deep Single Filtered Balance System, Stainless Steel, 4', 115 V 3252160 Mfr # 3971496
COMPARE
380 48 Tall & Deep Single Filtered Balance System, Stainless Steel, 4', 115 V
$25,813.79CAD / Each
Labconco RXPert  Tall & Deep Single Filtered Balance System, Epoxy Surface, 4', 115 V 3252162 Mfr # 3971497
COMPARE
380 48 Tall & Deep Single Filtered Balance System, Epoxy Surface, 4', 115 V
$24,421.44CAD / Each
Labconco RXPert  Tall Single Filtered Balance System, Stainless Steel Surface, 5', 115 V 3252166 Mfr # 3971576
COMPARE
480 60 Tall Single Filtered Balance System, Stainless Steel Surface, 5', 115 V
$30,978.02CAD / Each
Labconco RXPert  Tall Single Filtered Balance System, Epoxy Surface, 5', 115 V 3252168 Mfr # 3971577
COMPARE
480 60 Tall Single Filtered Balance System, Epoxy Surface, 5', 115 V
$30,027.69CAD / Each
Labconco RXPert  Tall & Deep Single Filtered Balance System, Stainless Steel, 5', 115 V 3252172 Mfr # 3971596
COMPARE
480 60 Tall & Deep Single Filtered Balance System, Stainless Steel, 5', 115 V
$37,888.22CAD / Each
Labconco RXPert  Tall & Deep Single Filtered Balance System, Epoxy Surface, 5', 115 V 3252174 Mfr # 3971597
COMPARE
480 60 Tall & Deep Single Filtered Balance System, Epoxy Surface, 5', 115 V
$38,897.50CAD / Each
Labconco RXPert  Tall Single Filtered Balance System, Stainless Steel Surface, 6', 115 V 3252178 Mfr # 3971676
COMPARE
580 72 Tall Single Filtered Balance System, Stainless Steel Surface, 6', 115 V
$32,746.09CAD / Each
Labconco RXPert  Tall Single Filtered Balance System, Epoxy Surface, 6', 115 V 3252180 Mfr # 3971677
COMPARE
580 72 Tall Single Filtered Balance System, Epoxy Surface, 6', 115 V
$32,547.19CAD / Each
Labconco RXPert  Tall & Deep Single Filtered Balance System, Stainless Steel, 6', 115 V 3252184 Mfr # 3971696
COMPARE
580 72 Tall & Deep Single Filtered Balance System, Stainless Steel, 6', 115 V
$39,641.56CAD / Each
Labconco RXPert  Tall & Deep Single Filtered Balance System, Epoxy Surface, 6', 115 V 3252186 Mfr # 3971697
COMPARE
580 72 Tall & Deep Single Filtered Balance System, Epoxy Surface, 6', 115 V
$41,203.35CAD / Each
Showing 1- 18 of 18