Labconco RXPert Tall & Deep Single Filtered Balance System, Stainless Steel, 6', 115 V

Representative image only.

Labconco RXPert Tall & Deep Single Filtered Balance System, Stainless Steel, 6', 115 V

Labconco | Mfr: 3971696| Item: 3252184

Keep hazardous powders and particulates contained

$39,641.56CAD / Each

$39,641.56CAD / Each

Buy It Together

Current Item

Labconco RXPert  Tall & Deep Single Filtered Balance System, Stainless Steel, 6', 115 V
Labconco RXPert Tall & Deep Single Filtered Balance System, Stainless Steel, 6', 115 V
3252184

$39,641.56CAD / Each
+ =
Total Price 49049.16 ADD 2 ITEMS TO CART
Product Description

Includes

8-ft (2.4-m) power cord.

Key Features

  • Meet USP <800> requirements for nonsterile hazardous compounding with redundant filtration optional
  • Bag in/bag out HEPA filter provides protection during filter changes
  • 1 year warranty

More About this Item

The RXPert cabinet features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement. The filtered balance system is a Class I enclosure that complies with USP requirements for nonsterile compounding.


All models include a built-in analog airflow monitor. Some models include a stainless steel or epoxy work surface; a work surface is required if one is not included.


Single-Filtered Enclosures include an all-in-one factory-installed canopy and the damper allows for thimble ducting to the outside. All models require a remote blower, ductwork, and supporting base (order separately).


Compliance: USP <800>, SEFA 1-2010, modified ASHRAE 110-1995, ANSI Z9.5-2011, UL 61010-1, and CAN/CSA C22.2 No. 61010.1 standards.

1 Year Warranty
Specifications
  • Hood Width (in)72
  • Exterior Width (in)72
  • Exterior Height (in)50.4375
  • Exterior Depth (in)35.5
  • Interior Width (in)69.625
  • Interior Height (in)32
  • Interior Depth (in)30
  • Power (VAC)115
  • Sash opening (in)12
  • Exhaust Volume (CFM)580
  • Shipping Weight (lbs)705
  • Shipping Weight (kg)320
  • DescriptionTall & Deep Single Filtered Balance System, Stainless Steel, 6', 115 V
  • Warranty1 Year
Reviews
Q&A