Labconco Protector Premier Fume Hoods
Excellent chemical resistance
Labconco Protector Premier Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general-purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
These by-pass hoods feature the largest unobstructed sightline available—37.5” from work surface to header panel. Other features include Eco-Foil™ air foil that corrects natural turbulence while keeping cords out of the way, dry powder-coated steel exterior, safety glass sash, and molded fiberglass exhaust collar.
Hoods are available with a built-in standard fiberglass blower, an explosion-proof fiberglass blower, or without a blower (remote blower sold separately). Standard hoods include T8 fluorescent lighting. Explosion-proof (EP) models include incandescent lighting (bulb not included). EP blowers are UL-listed for Class I, Group D and Class II, Groups E, F and G. All models are ETL listed. 230V models are CE listed.
Additional service fixtures can be added; contact a Cole-Parmer Application Specialist.
$16,339.90 - $32,805.03CAD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Power (VAC) Description
Availability Pricing
Labconco Protector Premier 4' Fume Hood With No Services, No Blower; 115V/60Hz 0910261 Mfr # 100400000
COMPARE
59 115 Fiberglass 48" Fume Hood, No Services, No Blower, 115 V, 60 Hz
$16,339.90CAD / Each
Labconco Protector Premier 4' Fume Hood With 2 Services, Electrical Duplex, No Blower; 115V/60Hz 0910262 Mfr # 100400002
COMPARE
59 115 Fiberglass 48" Fume Hood, Two Services and Duplex, No Blower, 115 V, 60 Hz
$18,476.31CAD / Each
Labconco Protector Premier 4' Fume Hood With Built-in Blower, No Services; 115V/60Hz 0910263 Mfr # 100400040
COMPARE
66 115 Fiberglass 48" Fume Hood, No Services, With Blower, 115 V, 60 Hz
$23,471.10CAD / Each
Labconco Protector Premier 4' Fume Hood With Two Services, Electrical Duplex, Built-in Blower; 115V/60Hz 0910264 Mfr # 100400042
COMPARE
66 115 Fiberglass 48" Fume Hood, Two Services and Duplex, With Blower, 115 V, 60 Hz
$23,264.83CAD / Each
Labconco Protector Premier 4' Fume Hood With Built-in Blower, Explosion Proof, No Services; 115V/60Hz 0910265 Mfr # 100400070
COMPARE
66 115 Fiberglass 48" Fume Hood, No Services, With Blower, Explosion Proof, 115 V, 60 Hz
$25,592.78CAD / Each
Labconco Protector Premier 4' Fume Hood With Built-in Blower, Explosion Proof, 2 Serivce; 115V/60Hz 0910266 Mfr # 100400071
COMPARE
66 115 Fiberglass 48" Fume Hood, with EP Blower, 2 Fixtures; 115 V, 60 Hz
$27,309.28CAD / Each
Labconco Protector Premier 5' Fume Hood With No Services, No Blower; 115V/60Hz 0910267 Mfr # 100500000
COMPARE
59 115 Fiberglass 60" Fume Hood, No Services, No Blower, 115 V, 60 Hz
$18,277.40CAD / Each
Labconco Protector Premier 5' Fume Hood With 2 Services, Electrical Duplex, No Built-in Blower; 115V/60Hz 0910268 Mfr # 100500002
COMPARE
59 115 Fiberglass 60" Fume Hood, Two Services and Duplex, No Blower, 115 V, 60 Hz
$20,435.92CAD / Each
Labconco Protector Premier 5' Fume Hood With Built-in Blower, No Services; 115V/60Hz 0910269 Mfr # 100500040
COMPARE
66 115 Fiberglass 60" Fume Hood, No Services, With Blower, 115 V, 60 Hz
$24,333.04CAD / Each
Labconco Protector Premier 5' Fume Hood, With  Built-in Blower, 2 Services, Electrical Duplex; 115V/60Hz 0910270 Mfr # 100500042
COMPARE
66 115 Fiberglass 60" Fume Hood, Two Services and Duplex, With Blower, 115 V, 60 Hz
$26,012.70CAD / Each
Labconco Protector Premier 5' Fume Hood, With Built-in Blower, Explosion Proof, No Services; 115V/60Hz 0910271 Mfr # 100500070
COMPARE
66 115 Fiberglass 60" Fume Hood, No Services, With Blower, Explosion Proof, 115 V, 60 Hz
$28,333.29CAD / Each
Labconco Protector Premier 5' Fume Hood With Built-in Blower, 2 Fixtures, Explosion Proof; 115V, 60Hz 0910272 Mfr # 100500071
COMPARE
66 115 Fiberglass 60" Fume Hood, with EP Blower, 2 Fixtures; 115 V, 60 Hz
$30,064.52CAD / Each
Labconco Protector Premier 6' Fume Hood With No Services, No Blower; 115V/60Hz 0910273 Mfr # 100600000
COMPARE
59 115 Fiberglass 72" Fume Hood, No Services, No Blower, 115 V, 60 Hz
$20,229.64CAD / Each
Labconco Protector Premier 6' Fume Hood With 2 Services and Duplex, No Blower; 115V/60Hz 0910274 Mfr # 100600002
COMPARE
59 115 Fiberglass 72" Fume Hood, Two Services and Duplex, No Blower, 115 V, 60 Hz
$22,373.43CAD / Each
Labconco Protector Premier 6' Fume Hood With No Services, With Built-in Blower; 115V/60Hz 0910275 Mfr # 100600040
COMPARE
66 115 Fiberglass 72" Fume Hood, No Services, With Blower, 115 V, 60 Hz
$26,609.42CAD / Each
Labconco Protector Premier 6' Fume Hood  With Built-in Blower, 2 Services, Electrical Duplex; 115V/60Hz 0910276 Mfr # 100600042
COMPARE
66 115 Fiberglass 72" Fume Hood, Two Services and Duplex, With Blower, 115 V, 60 Hz
$28,745.84CAD / Each
Labconco Protector Premier 6' Fume Hood With Built-in Blower, Explosion Proof, No Service; 115V/60Hz 0910277 Mfr # 100600070
COMPARE
66 115 Fiberglass 72" Fume Hood, No Services, With Blower, Explosion Proof, 115 V, 60 Hz
$31,103.26CAD / Each
Labconco Protector Premier 6' Fume Hood With Built-in Blower, 2 Services, Explosion Proof; 115V/60Hz 0910278 Mfr # 100600071
COMPARE
66 115 Fiberglass 72" Fume Hood, Two Services, With Blower, Explosion Proof, 115 V, 60 Hz
$32,805.03CAD / Each
Labconco Protector Premier 4' Fume Hood With Built-in Blower, 2 Services; 230V/60Hz 0910282 Mfr # 100400061
COMPARE
66 230 Fiberglass 48" Fume Hood, Two Services, With Blower, 230 V, 50 Hz
$22,844.91CAD / Each
Labconco Protector Premier 5' Fume Hood With Built-in Blower, No Services; 230V/60Hz 0910285 Mfr # 100500060
COMPARE
66 230 Fiberglass 60" Fume Hood, No Services, With Blower, 230 V, 50 Hz
$23,854.18CAD / Each
Showing 1- 20 of 20