Labconco Protector Premier 6' Fume Hood With No Services, With Built-in Blower; 115V/60Hz

Representative image only.

Labconco Protector Premier 6' Fume Hood With No Services, With Built-in Blower; 115V/60Hz

Labconco | Mfr: 100600040| Item: 0910275

Excellent chemical resistance

$26,609.42CAD / Each

$26,609.42CAD / Each

Buy It Together

Current Item

Labconco Protector Premier 6' Fume Hood With No Services, With Built-in Blower; 115V/60Hz
Labconco Protector Premier 6' Fume Hood With No Services, With Built-in Blower; 115V/60Hz
0910275

$26,609.42CAD / Each
+ + + + + =
Total Price 95659.84 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Labconco Protector Premier Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general-purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.

These by-pass hoods feature the largest unobstructed sightline available—37.5” from work surface to header panel. Other features include Eco-Foil™ air foil that corrects natural turbulence while keeping cords out of the way, dry powder-coated steel exterior, safety glass sash, and molded fiberglass exhaust collar.

Hoods are available with a built-in standard fiberglass blower, an explosion-proof fiberglass blower, or without a blower (remote blower sold separately). Standard hoods include T8 fluorescent lighting. Explosion-proof (EP) models include incandescent lighting (bulb not included). EP blowers are UL-listed for Class I, Group D and Class II, Groups E, F and G. All models are ETL listed. 230V models are CE listed.

Additional service fixtures can be added; contact a Cole-Parmer Application Specialist.

ETL Listed
Specifications
  • Hood Width (in)72
  • Blower TypeStandard
  • Blower (hp)1/2
  • LightsFluorescent
  • Switches2
  • Exterior Width (in)72
  • Exterior Height (in)66
  • Exterior Depth (in)31.6875
  • Power (VAC)115
  • Power (Hz)60
  • Sash opening (in)37.5
  • Shipping Weight (lbs)495
  • Shipping Weight (kg)225
  • Ductwork Diameter (in)12
  • DescriptionFiberglass 72" Fume Hood, No Services, With Blower, 115 V, 60 Hz
Reviews
Q&A