Labconco® Protector® Echo™ Ductless Fume Hoods
Combine Neutrodine® Unisorb technology for simultaneous capture of solvents, acids, and bases with eco-friendly DH III protection
  • Neutrodine® Unisorb molecular carbon filter offers comprehensive molecular filtration
  • eGuard® communication software offers real-time monitoring from remote locations
  • Constructed of durable white powder-coated steel exterior with a chemical-resistant, fiberglass-reinforced, composite panel liner
  • Ensure safety with built-in sensors to alert to primary filter breakthrough of solvent and acid fumes, laboratory air quality, sash position, and temperature
  • DH III filtration setup ensures safety with backup a filter that has the same retention potential as the main filter
The Labconco® Protector® Echo™ ductless fume hoods offer the safety of Neutrodine® Unisorb filtration technology. The Neutrodine® Unisorb modules allow for simultaneous capture of solvents, acids, and bases with an additional level of safety preventing release of pollutants into the lab. All models feature a single piece, tempered safety glass vertical-rising sash with cable pulley, sash weight and powder-coated aluminum sash handle, and tracks with airflow openings.
The Echo™ fume hoods also include pre-wired vapor-proof LED lights for up to 63 foot candles, built-in exhaust fans to maintain 60 fpm face velocity at 16" sash opening height, and a smart command control module. This control module allows you to control fans and lights while also monitoring performance through visual and audible signals. Alarms include a maintenance alarm, filter status, fan status, and face velocity alarm. Easily connect to a Smartphone or computer via Ethernet and Wi-Fi. Fume hoods also include eGuard® communication software for real-time remote monitoring of data such as usage authorization, filter saturation detection, sash position, and temperature.
Choose from models with two pre-plumed service fixtures with forged brass valves for gas and cold water or pre-wired GFCI electrical duplex receptacle. Model options also include hoods without windows, hoods with side windows, or hoods with side and back windows. All models require, but do not include, Neutrodine® Unisorb filters, work surface, and base cabinet or stand. Order an optional HEPA filter for 99.995% (H14) high efficiency particulate air filtration for use with particulate-containing applications or when installed in cleanrooms.
$40,739.23 - $59,635.46CAD / Each
60 variations of this product are available.
See All Product Options Below
60 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fixtures Hood Width (in) Power (VAC)
Availability Pricing
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 48" W, No Windows; 115V, 50/60 Hz 2861601 Mfr # 180410400
COMPARE
No fixture or GFCI Duplex 48 115
$40,739.23CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 48" W, No Windows, 2 Fixtures; 115V, 50/60 Hz 2861603 Mfr # 180410401
COMPARE
Two fixtures and no GFCI Duplex 48 115
$42,205.26CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 48" W, No Windows, 2 Fixtures and GFCI Duplex; 115V, 50/60 Hz 2861605 Mfr # 180410402
COMPARE
Two fixtures and one GFCI Duplex 48 115
$42,411.53CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 48" W, No Windows; 230V, 50/60 Hz 2861607 Mfr # 180410420
COMPARE
No fixture or GFCI Duplex 48 230
$40,739.23CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 48" W, No Windows, 2 Fixtures; 230V, 50/60 Hz 2861609 Mfr # 180410421
COMPARE
Two fixtures and no GFCI Duplex 48 230
$42,205.26CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 60" W, No Windows; 115V, 50/60 Hz 2861611 Mfr # 180510400
COMPARE
No fixture or GFCI Duplex 60 115
$43,604.98CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 60" W, No Windows, 2 Fixtures; 115V, 50/60 Hz 2861613 Mfr # 180510401
COMPARE
Two fixtures and no GFCI Duplex 60 115
$45,085.73CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 60" W, No Windows, 2 Fixtures and GFCI Duplex; 115V, 50/60 Hz 2861615 Mfr # 180510402
COMPARE
Two fixtures and one GFCI Duplex 60 115
$45,284.64CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 60" W, No Windows; 230V, 50/60 Hz 2861617 Mfr # 180510420
COMPARE
No fixture or GFCI Duplex 60 230
$43,604.98CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 60" W, No Windows, 2 Fixtures; 230V, 50/60 Hz 2861619 Mfr # 180510421
COMPARE
Two fixtures and no GFCI Duplex 60 230
$45,085.73CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 72" W, No Windows; 115V, 50/60 Hz 2861621 Mfr # 180610400
COMPARE
No fixture or GFCI Duplex 72 115
$46,455.99CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 72" W, No Windows, 2 Fixtures; 115V, 50/60 Hz 2861623 Mfr # 180610401
COMPARE
Two fixtures and no GFCI Duplex 72 115
$47,936.74CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 72" W, No Windows, 2 Fixtures and GFCI Duplex; 115V, 50/60 Hz 2861625 Mfr # 180610402
COMPARE
Two fixtures and one GFCI Duplex 72 115
$48,143.02CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 72" W, No Windows; 230V, 50/60 Hz 2861627 Mfr # 180610420
COMPARE
No fixture or GFCI Duplex 72 230
$46,455.99CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 72" W, No Windows, 2 Fixtures; 230V, 50/60 Hz 2861628 Mfr # 180610421
COMPARE
Two fixtures and no GFCI Duplex 72 230
$47,936.74CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 96" W, No Windows; 115V, 50/60 Hz 2861629 Mfr # 180810400
COMPARE
No fixture or GFCI Duplex 96 115
$54,110.25CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 96" W, No Windows, 2 Fixtures; 115V, 50/60 Hz 2861630 Mfr # 180810401
COMPARE
Two fixtures and no GFCI Duplex 96 115
$55,598.37CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 96" W, No Windows, 2 Fixtures and 2 GFCI Duplexes; 115V, 50/60 Hz 2861631 Mfr # 180810402
COMPARE
Two fixtures and two GFCI Duplexes 96 115
$55,789.91CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 96" W, No Windows; 230V, 50/60 Hz 2861633 Mfr # 180810420
COMPARE
No fixture or GFCI Duplex 96 230
$54,110.25CAD / Each
Labconco Protector® Echo™ Ductless Fume Hood with Solvent and Acid Sensors, 96" W, No Windows, 2 Fixtures; 230V, 50/60 Hz 2861635 Mfr # 180810421
COMPARE
Two fixtures and no GFCI Duplex 96 230
$55,598.37CAD / Each
Showing 1 - 20 of 60