Labconco Fiberglass 30 Laboratory Hoods
Small fume hoods with big benefits
These 30-inch fiberglass fume hoods save space while still offering many of the features and benefits of the larger hoods. Use these efficient hoods for applications requiring small work space and good fume removal.
One-piece fiberglass interior gives this hood outstanding chemical- and fire-resistance. Pre-set baffle and by-pass airflow design assure consistent ventilation regardless of sash position. The steel-constructed exterior is glacier white epoxy-coated. Hoods feature a vertical-rising tempered glass safety sash with epoxy-coated aluminum handle.
Available with or without a built-in blower, in standard or spark-proof configurations. Hoods requires a work surface--order separately. Complete your fume hood with a variety of accessories, including a storage cabinet and an air monitor (order separately). Call our Applications Specialists regarding blower sizing and ordering service fixtures, electrical duplexes, and other custom features.
What’s included:All models are shipped fully assembled. Hard-wired installations.
$10,402.13 - $13,606.76CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Power (VAC) Description
Availability Pricing
Labconco 3030002 Fiberglass 30 fume hood, no blower, 115V 0906610 Mfr # 3030002
COMPARE
48 115 Fiberglass 30" Fume Hood, No Blower, 115 V
$10,402.13CAD / Each
Labconco 3030000 Fiberglass 30 fume hood, 1/10 HP blower, 115V 0906630 Mfr # 3030000
COMPARE
48 110 Fiberglass 30" Fume Hood, 1/10 HP Blower, 115 V
$11,853.42CAD / Each
Labconco 3030004 Fiberglass 30 fume hood, explosion-proof, 1/4 HP blower, no lamp, 115V 0906650 Mfr # 3030004
COMPARE
54 115 Fiberglass 30" Fume Hood, Explosion-Proof, 1/4 HP Blower, No Lamp, 115 V
$13,606.76CAD / Each