Lab Armor Waterless Bead Baths
Low maintenance, eco-friendly alternative to water baths
  • Eco-friendly — beads are recyclable, use 50% less electricity, and do not require germicides
  • Beads hold things in place, eliminating the need for racks and weights
  • Bath is always ready, eliminating warm-up times
  • Safer operation than oil baths
  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
These baths use metallic beads instead of water, delivering exceptional temperature uniformity of +/-1 degree without lengthy warm-up times. Beads create a dry bath, avoiding the contamination and maintenance of traditional water baths. They also improve energy efficiency over 50% of traditional water baths.
Beads hold a variety of containers securely in place without any float-away or ruined samples. You are no longer limited to water-tight containers, so you can incubate multi-well plates, petri dishes, and open-top samples at any angle.
Metallic beads have high thermal conductivity in the range of +/-180° C. Smooth, rounded surfaces measure 5 to 8 mm diameter and 1 to 2 mm high. Beads are moisture and gas impermeable, and are easy to keep clean without harmful germicides. These versatile beads can also be used in a container for freezers, ovens, and incubators.
$3,646.64 - $4,869.55CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L) Description
Availability Pricing
Lab Armor 74300-706 Waterless Bead Bath without Beads, 6 L Capacity; 120 VAC 1012202 Mfr # 74300-706
COMPARE
115 6 Waterless Bead Bath without Beads, 6 L Capacity; 120 VAC
$4,169.69CAD / Each
Lab Armor 74200-706 Waterless Bead Bath without Beads, 6 L Capacity; 220 VAC 1012207 Mfr # 74200-706
COMPARE
230 6 Waterless Bead Bath without Beads, 6 L Capacity; 220 VAC
$4,869.55CAD / Each
Lab Armor 74300-714 Waterless Bead Bath without Beads, 14 L Capacity; 120 VAC 1012212 Mfr # 74300-714
COMPARE
115 14 Waterless Bead Bath without Beads, 14 L Capacity; 120 VAC
$3,646.64CAD / Each
Lab Armor 74200-714 Waterless Bead Bath without Beads, 14 L Capacity; 220 VAC 1012217 Mfr # 74200-714
COMPARE
230 14 Waterless Bead Bath without Beads, 14 L Capacity; 220 VAC
$3,757.14CAD / Each
Lab Armor 74300-720 Waterless Bead Bath without Beads, 20 L Capacity; 120 VAC 1012222 Mfr # 74300-720
COMPARE
115 20 Waterless Bead Bath without Beads, 20 L Capacity; 120 VAC
$4,825.35CAD / Each
Lab Armor 74200-720 Waterless Bead Bath without Beads, 20 L Capacity; 220 VAC 1012227 Mfr # 74200-720
COMPARE
230 20 Waterless Bead Bath without Beads, 20 L Capacity; 220 VAC
$4,766.42CAD / Each