Lab Armor 74200-720 Waterless Bead Bath without Beads, 20 L Capacity; 220 VAC

Representative image only.

Lab Armor 74200-720 Waterless Bead Bath without Beads, 20 L Capacity; 220 VAC

Lab Armor | Mfr: 74200-720| Item: 1012227

Low maintenance, eco-friendly alternative to water baths

$4,766.42CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$4,766.42CAD / Each

Buy It Together

Current Item

Lab Armor 74200-720 Waterless Bead Bath without Beads, 20 L Capacity; 220 VAC
Lab Armor 74200-720 Waterless Bead Bath without Beads, 20 L Capacity; 220 VAC
1012227

$4,766.42CAD / Each
+
Metallic Bath Beads for Dry Baths; 2-Liters
0627450

$649.03CAD / Each
+
Cole-Parmer WB-200 Water Bath, 11 L; 120 VAC, 60 Hz
7890522

$1,119.78CAD / Each
+ =
Total Price 7382.43 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

stainless steel lid and power cord. Purchase bath with beads or bath only and beads separately.

Key Features

  • Eco-friendly — beads are recyclable, use 50% less electricity, and do not require germicides
  • Beads hold things in place, eliminating the need for racks and weights
  • Bath is always ready, eliminating warm-up times
  • Safer operation than oil baths
  • Dry, inert, nontoxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
  • 1 year warranty

More About this Item

These baths use metallic beads instead of water, delivering exceptional temperature uniformity of +/-1 degree without lengthy warm-up times. Beads create a dry bath, avoiding the contamination and maintenance of traditional water baths. They also improve energy efficiency over 50% of traditional water baths.

Beads hold a variety of containers securely in place without any float-away or ruined samples. You are no longer limited to water-tight containers, so you can incubate multi-well plates, petri dishes, and open-top samples at any angle.

Metallic beads have high thermal conductivity in the range of +/-180° C. Smooth, rounded surfaces measure 5 to 8 mm diameter and 1 to 2 mm high. Beads are moisture and gas impermeable, and are easy to keep clean without harmful germicides. These versatile beads can also be used in a container for freezers, ovens, and incubators.

Order beads separately. (16L to bath fill line).

Conformite Europeenne1 Year Warranty
Specifications
  • Bath TypeBead Bath
  • Reservoir Capacity (L)20
  • Max Temperature (° C)80
  • Min Temperature (° C)Ambient +5
  • Temperature Uniformity (° C)±1.0
  • Width (in)15 7/8
  • Height (in)23 3/8
  • Depth (in)8 1/2
  • Bath Opening Width (InchShort)6
  • Bath Opening Length (in)17 1/8
  • Immersion Depth (in)6
  • Temperature SettingDigital
  • Wetted MaterialsSS
  • Power (VAC)230
  • Power (Hz)50/60
  • Power (amps)3
  • DescriptionWaterless Bead Bath without Beads, 20 L Capacity; 220 VAC
  • Warranty1 Year
Reviews
Q&A