KNF Laboport Filtration Pumps
Great for vacuum filtration, blotting, and solid phase extraction!
  • Lightweight but rugged design minimizes downtime for repairs
  • No wetted metal parts in the pump head for corrosion protection
  • Corrosion resistant materials to meet demanding applications
An exclusive multiport valve design tolerates occasional liquids drawn into the vacuum system. All wetted materials are made of chemical resistant resins. Typical applications include bottle-top filtration, manifold filtration, solvent degassing, and desiccation.
$1,824.06 - $3,368.17CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg) Motor hp
Availability Pricing
KNF N816.3 KTP Vacuum/pressure Pump, 16 L/min, 15 Torr, 7.4 PSIG, 115 VAC 7816446 Mfr # 057013/055162
COMPARE
0.6 29.4 1/8
$2,728.72CAD / Each
KNF N816.1.2KTP Vacuum/pressure Pump, 30 L/min, 120 Torr, 7.4 PSIG, 115 VAC 7816448 Mfr # 057017/055162
COMPARE
1.1 25.3 1/8
$2,687.46CAD / Each
KNF N811 KV.45P Filtration Pump, Gauge/Reg, PTFE/PPS/FKM; 0.45cfm/27"Hg-30psi/115V 7816462 Mfr # 077408/075655
COMPARE
0.45 29.4 1/10
$1,824.06CAD / Each
KNF N816.3KT345P Filtration Pump, Gauge/Reg, PTFE/PPS/FFKM; 0.6cfm/29.4"Hg-7.4psi/115V 7816464 Mfr # 077271/000000
COMPARE
0.6 29.4 1/12
$3,368.17CAD / Each