Kimble Glass Desiccator
Durable and efficient for daily use
  • Conforms to USP Type III and ASTM E438, Type II requirements
Soda-lime glass desiccators include a metal plate, s solid lid with a threaded plastic top knob and a recessed base for containment of desiccant. Excellent for storage of hygroscopic chemicals, and to prevent absorption of moisture when using crucibles, weighing bottles, and watch glasses.
$679.97 - $1,242.07CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (InchShort) Material Plate Size (mm)
Availability Pricing
DWK Life Sciences (Kimble) Glass Desiccator with Plastic top Knob, 150 mm 0653641 Mfr # 31050-150
COMPARE
10 Glass 150
$679.97CAD / Each
DWK Life Sciences (Kimble) Glass Desiccator with Plastic top Knob, 200 mm 0653651 Mfr # 31050-200
COMPARE
12.25 Glass 200
$1,055.68CAD / Each
DWK Life Sciences (Kimble) Glass Desiccator with Plastic top Knob, 250 mm 0653661 Mfr # 31050-250
COMPARE
13 Glass 250
$1,242.07CAD / Each