KIMAX Glass Vacuum Desiccators
Greased collar valve allows for better grip and greater degree of durability
  • Maximum vacuum of 20" Hg
Manufactured from soda-lime glass these vacuum desiccators include a lid with collar valve, metal plate, and a recessed base for desiccant containment. Conform to USP Type III and ASTM E438, Type II requirements. Valve stem accepts 5/16" ID flexible tubing.
$1,024.01 - $1,688.50CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (InchShort) Material Plate Size (mm)
Availability Pricing
DWK Life Sciences (Kimble) Glass Vacuum Desiccator with Collar, 150 mm 0653611 Mfr # 31200-150
COMPARE
10 Glass 150
$1,024.01CAD / Each
DWK Life Sciences (Kimble) Glass Vacuum Desiccator with Collar, 200 mm 0653621 Mfr # 31200-200
COMPARE
12.25 Glass 200
$1,435.82CAD / Each
DWK Life Sciences (Kimble) Glass Vacuum Desiccator with Collar, 250 mm 0653631 Mfr # 31200-250
COMPARE
13 Glass 250
$1,688.50CAD / Each