Julabo Open Bath Tanks
Large generous opening provides easy access to samples
The Julabo open bath tanks, combined with the Corio™ heating immersion circulators, are ideally suited for controlling liquid media temperature. Generous bath openings provide ready access to samples. Choose from two different tank materials—the transparent polycarbonate baths allow for heating up to 100°C while the stainless steel baths can go up to 150°C.
$892.87 - $6,183.82CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Reservoir Capacity (L) Wetted Materials
Availability Pricing
Julabo CORIO™ BT9 Bath Tank, Plastic; 9 L 1214702 Mfr # 9901309
COMPARE
100 9 Polycarbonate
$892.87CAD / Each
Julabo CORIO™ BT19 Bath Tank, Plastic; 19 L 1214703 Mfr # 9901319
COMPARE
100 19 Polycarbonate
$1,162.50CAD / Each
Julabo CORIO™ BT27 Bath Tank, Plastic; 27 L 1214704 Mfr # 9901327
COMPARE
100 27 Polycarbonate
$1,331.94CAD / Each
Julabo CORIO™ B5 Bath Tank, Stainless Steel; 5 L 1214706 Mfr # 9903405
COMPARE
150 5 Stainless steel
$988.64CAD / Each
Julabo CORIO™ B13 Bath Tank, Stainless Steel; 13 L 1214707 Mfr # 9903413
COMPARE
150 13 Stainless steel
$1,695.87CAD / Each
Julabo CORIO™ B17 Bath Tank, Stainless Steel; 17 L 1214708 Mfr # 9903417
COMPARE
150 17 Stainless steel
$1,990.55CAD / Each
Julabo CORIO™ B27 Bath Tank, Stainless Steel; 27 L 1214711 Mfr # 9903427
COMPARE
150 27 Stainless steel
$2,538.65CAD / Each
Julabo CORIO™ B33 Bath Tank, Stainless Steel; 33 L 1214712 Mfr # 9903433
COMPARE
150 33 Stainless steel
$5,865.57CAD / Each
Julabo CORIO™ B39 Bath Tank, Stainless Steel; 39 L 1214713 Mfr # 9903439
COMPARE
150 39 Stainless steel
$6,183.82CAD / Each
Showing 1- 9 of 9