Julabo MAGIO™ MS / MX Bridge-Mounted Circulators
Extremely powerful pump with adjustable bridge
  • Performance values up to 31 L/min or 0.92 bar (pressure)/-0.4 bar (suction)
  • All wetted parts are constructed of stainless steel for chemical resistance
  • High-resolution, TFT touch-screen display features a multi-lingual user interface
  • Remote control and data management made easy with USB interface, RS-232, and Ethernet
  • Pt100 external sensor connection for measurement and control
MAGIO™ bridge-mounted circulators combine high temperature control performance with maximum flexibility. The adjustable bridge means that the circulators can be used with bath tanks up to a filling volume of 100 liters.
$11,106.41 - $11,477.71CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Immersion Depth (in) Power (VAC)
Availability Pricing
Julabo MS-Z Bridge Mounted Heating Circulator (Immersion Depth of 150 mm | 5.9") for Tanks to 100 L with 1 kW (115V) Heating Power to +300 °C RK-12152-80 Mfr # 9032201.22
COMPARE
5 29/32 115
$11,106.41CAD / Each
Julabo MS-Z Bridge Mounted Heating Circulator (Immersion Depth of 150 mm | 5.9") for Tanks to 100 L with 2 kW (230V) Heating Power to +300 °C RK-12152-81 Mfr # 9032201.33
COMPARE
5 29/32 230
$11,106.41CAD / Each
Julabo MX-Z Bridge Mounted Heating Circulator (Immersion Depth of 200 mm | 7.9") for Tanks to 100 L with 3 kW (230V) Heating Power to +300 °C RK-12152-82 Mfr # 9033201.33
COMPARE
7 29/32 230
$11,477.71CAD / Each