Julabo FT Series Immersion Chillers with Probe
Economical alternative to dry ice or tap water
  • Compact size maximizes bench space
  • Immersion probe rapidly cools liquids
  • Designed for continuous operation
These compact immersion chillers offer an ideal solution to rapidly cooling liquids down to ambient temperature or below. Great for use with heating circulators. Featuring an immersion cooling probe, these chillers are easy to operate and manage a variety of cooling applications. Carrying handles and flexible hose make it easy to move the chillers around and reposition as needed.
$8,728.36 - $27,593.65CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 20° C (watts) Min Temperature (° C) Power (VAC)
Availability Pricing
Julabo FT200 Immersion Chiller, -20 to 30°C, 115V/60Hz 1215051 Mfr # 9650820.2
COMPARE
250 -20 115
$8,728.36CAD / Each
Julabo FT900 Immersion Chiller, -90 to 30°C, 115V/60Hz 1215057 Mfr # 9650890.2
COMPARE
300 -90 115
$27,593.65CAD / Each
Julabo FT900 Immersion Chiller, -90 to 30°C, 230V/50-60Hz 1215058 Mfr # 9650890.3
COMPARE
300 -90 230
$27,593.65CAD / Each