Julabo CORIO™ Open Heating Circulating Baths
Easily view bright white LED display from across the room
  • Remains stable at ambient temperatures of up to 40°C
  • Securely locks in power cord for safe connection
These open heating circulating baths provide a wide variety of temperature applications up to 150°C. The baths come in two different tank materials—either high-quality transparent polycarbonate or stainless steel with volumes up to 39 L. The transparent heating bath circulators feature an adjustable jet nozzle while the stainless steel baths offer an integrated drain screw at the front for easy removal of bath fluids (except the 5-L models). The bright digital display and intuitive operating controls and safety functions allow for quick, simple operation. The circulator can be removed for easy cleaning of the bath tank.
Basic open heating circulating baths are designed specifically for internal applications. Common applications include temperature control of samples, preparation of samples for serology and clinical chemistry, analysis, and material testing.
Advanced open heating circulating baths are designed for internal and external temperature applications. They include the same features as the basic models plus a USB interface for remote control and transfer of data to a PC, pump control level to easily change between internal and external circulation, and inclined pump connections (M16x1) to facilitate the connection of applications. Safety features include automatic high temperature shut off and low liquid level alarm. Common applications include temperature control of measurement cells, photometers, and samples; heating of glass flasks; and temperature testing of food, luxury articles, and materials.
$3,260.61 - $6,257.49CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
Julabo CORIO™ C-BT5 Heating Circulating Bath, 1 kW, 5 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz 1214811 Mfr # 9011305.22
COMPARE
100 100 to 115 Heating Circulating Bath, 1 kW, 5 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz
$3,260.61CAD / Each
Julabo CORIO™ C-BT9 Heating Circulating Bath, 1 kW, 9 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz 1214815 Mfr # 9011309.22
COMPARE
100 100 to 115 Heating Circulating Bath, 1 kW, 9 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz
$3,421.21CAD / Each
Julabo CORIO™ C-BT9 Heating Circulating Bath, 2 kW, 9 L Plastic Tank, 20-100°C; 230 V/50-60 Hz 1214817 Mfr # 9011309.33
COMPARE
100 230 Heating Circulating Bath, 2 kW, 9 L Plastic Tank, 20-100°C; 230 V/50-60 Hz
$3,421.21CAD / Each
Julabo CORIO™ C-BT19 Heating Circulating Bath, 1 kW, 19 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz 1214819 Mfr # 9011319.22
COMPARE
100 100 to 115 Heating Circulating Bath, 1 kW, 19 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz
$3,658.43CAD / Each
Julabo CORIO™ C-BT27 Heating Circulating Bath, 1 kW, 27 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz 1214823 Mfr # 9011327.22
COMPARE
100 100 to 115 Heating Circulating Bath, 1 kW, 27 L Plastic Tank, 20-100°C; 100-115 V/50-60 Hz
$3,805.77CAD / Each
Julabo CORIO™ C-BT27 Heating Circulating Bath, 2 kW, 27 L Plastic Tank, 20-100°C; 230 V/50-60 Hz 1214825 Mfr # 9011327.33
COMPARE
100 230 Heating Circulating Bath, 2 kW, 27 L Plastic Tank, 20-100°C; 230 V/50-60 Hz
$3,805.77CAD / Each
Julabo CORIO™ C-B5 Heating Circulating Bath, 1 kW, 5 L SS Tank, 20-100°C; 100-115 V/50-60 Hz 1214827 Mfr # 9011405.22
COMPARE
100 100 to 115 Heating Circulating Bath, 1 kW, 5 L SS Tank, 20-100°C; 100-115 V/50-60 Hz
$3,539.08CAD / Each
Julabo CORIO™ C-B5 Heating Circulating Bath, 2 kW, 5 L SS Tank, 20-100°C; 230 V/50-60 Hz 1214829 Mfr # 9011405.33
COMPARE
100 230 Heating Circulating Bath, 2 kW, 5 L SS Tank, 20-100°C; 230 V/50-60 Hz
$3,539.08CAD / Each
Julabo CORIO™ C-B19 Heating Circulating Bath, 1 kW, 19 L SS Tank, 20-100°C; 100-115 V/50-60 Hz 1214837 Mfr # 9011419.22
COMPARE
100 100 to 115 Heating Circulating Bath, 1 kW, 19 L SS Tank, 20-100°C; 100-115 V/50-60 Hz
$4,427.54CAD / Each
Julabo CORIO™ C-B27 Heating Circulating Bath, 1 kW, 27 L SS Tank, 20-100°C; 100-115 V/50-60 Hz 1214839 Mfr # 9011427.22
COMPARE
100 100 to 115 Heating Circulating Bath, 1 kW, 27 L SS Tank, 20-100°C; 100-115 V/50-60 Hz
$4,928.49CAD / Each
Julabo CORIO™ CD-BT5 Heating Circulating Bath, 1 kW, 5 L Plastic Tank, 20-100°C, Adj. Pump, USB; 115 V/60 Hz 1214841 Mfr # 9012305.02
COMPARE
100 115 Heating Circulating Bath, 1 kW, 5 L Plastic Tank, 20-100°C, Adj. Pump, USB; 115 V/60 Hz
$4,508.57CAD / Each
Julabo CORIO™ CD-BT19 Heating Circulating Bath, 1 kW, 19 L Plastic Tank, 20-100°C, Adj. Pump, USB; 115 V/60 Hz 1214844 Mfr # 9012319.02
COMPARE
100 115 Heating Circulating Bath, 1 kW, 19 L Plastic Tank, 20-100°C, Adj. Pump, USB; 115 V/60 Hz
$4,987.43CAD / Each
Julabo CORIO™ CD-B13 Heating Circulating Bath, 1 kW, 13 L SS Tank, 20-150°C, Adj. Pump, USB; 115 V/60 Hz 1214853 Mfr # 9012413.02
COMPARE
150 115 Heating Circulating Bath, 1 kW, 13 L SS Tank, 20-150°C, Adj. Pump, USB; 115 V/60 Hz
$5,514.90CAD / Each
Julabo CORIO™ CD-B19 Heating Circulating Bath, 1 kW, 19 L SS Tank, 20-150°C, Adj. Pump, USB; 115 V/60 Hz 1214859 Mfr # 9012419.02
COMPARE
150 115 Heating Circulating Bath, 1 kW, 19 L SS Tank, 20-150°C, Adj. Pump, USB; 115 V/60 Hz
$5,758.01CAD / Each
Julabo CORIO™ CD-B27 Heating Circulating Bath, 2 kW, 27 L SS Tank, 20-150°C, Adj. Pump, USB, 230 V/60 Hz 1214864 Mfr # 9012427.13
COMPARE
150 230 Heating Circulating Bath, 2 kW, 27 L SS Tank, 20-150°C, Adj. Pump, USB, 230 V/60 Hz
$6,257.49CAD / Each
Showing 1- 15 of 15