Julabo CORIO™ Heating Immersion Circulators
Convert any tank into a heated circulating bath with precise temperature control
  • Easily view bright white digital readings from across the room
  • Attach quickly to existing baths with universal clamp
  • Automatic high temperature and low liquid level shutoff optimize safety
The CORIO™ heating immersion circulators are ideal for precise temperature control of samples. A universal mounting clamp easily attaches the circulator onto an existing straight or round bath tank with a wall thickness up to 26 mm. All operating controls and safety functions are easily accessible from the front of unit, while the bright LED display allows temperature reading to be viewed from across the room. Other features include an adjustable pump nozzle, automatic high temperature shutoff, low liquid level alarm, and lockable power cord.
Choose from three CORIO™ models—C, CD, or CP. CORIO C is the basic model for internal standard applications for tanks up to 30L, no interface control, and a maximum working temperature of 100 °C. CORIO CD has all the advantages of the CORIO C plus a pump for internal and external applications, USB interface, and a maximum working temperature of 150 °C. CORIO CP has all the same features of the CORIO CD, with a pump for internal and external applications, plus USB and RS-232 interfaces, and a maximum working temperature of 200 °C.
$2,624.11 - $5,260.00CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Interface Max Temperature (° C) Power (VAC)
Availability Pricing
Julabo CORIO™ C Heating Immersion Circulator, 20 to 100 °C, Up to 30 L, 1 kW; 100-115 VAC, 50/60 Hz 1214801 Mfr # 9011000.22
COMPARE
None 100 100 to 115
$2,624.11CAD / Each
Julabo CORIO™ C Heating Immersion Circulator, 20 to 100 °C, Up to 30 L, 2 kW; 230 VAC, 50/60 Hz 1214803 Mfr # 9011000.33
COMPARE
None 100 230
$2,624.11CAD / Each
Julabo CORIO™ CD Heating Immersion Circulator, 20 to 150 °C, Up to 50 L, USB, 1 kW; 115 VAC, 60 Hz 1214805 Mfr # 9012000.02
COMPARE
USB 150 100 to 115
$3,036.66CAD / Each
Julabo CORIO™ CD Heating Immersion Circulator, 20 to 150 °C, Up to 50 L, USB, 2 kW; 230 VAC, 60 Hz 1214807 Mfr # 9012001.13
COMPARE
USB 150 230
$2,644.74CAD / Each
Julabo CORIO™ CP Heating Immersion Circulator, 20 to 200 °C, Up to 50 L, USB and RS-232, 1 kW; 100-115 VAC, 50/60 Hz 1215399 Mfr # 9013000.22
COMPARE
USB and RS-232 200 100 to 115
$5,260.00CAD / Each
Julabo CORIO™ CP Heating Immersion Circulator, 20 to 200 °C, Up to 50 L, USB and RS-232, 2 kW; 200-230 VAC, 50/60 Hz 1215400 Mfr # 9013000.33
COMPARE
USB and RS-232 200 200 / 230
$5,260.00CAD / Each