Julabo CORIO™ CP Refrigerating/Heating Circulators
Space-saving cooling coil provides more usable space
  • Easy pump change-over from internal to external circulation
  • Easily view bright white LED display from across the room
  • Remote control and data management made easy with USB and RS-232 interfaces
  • Meets Class III (FL) according to DIN 12876-1
The CORIO™ CP refrigerated/heating circulators can easily be switched between internal and external circulation. All operating controls and safety functions can be easily accessed from the front of unit. One power switch controls the circulator and refrigeration unit while the pump control lever, located below the bright digital LED display, makes changing between internal and external circulation simple. Inclined pump connections (M16x1) facilitate the connection of external applications.
Designed with a space-saving cooling coil provides more usable space in the bath tank. Other features include a front-mounted drain tap for removing bath fluids without the use of tools, USB and RS-232 interfaces for remote control and data management, removable front magnetic grid for easy cleaning and maintenance, lockable power plug, and an early warning system for low liquid level.
$13,496.25 - $23,921.96CAD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 20° C (watts) Power (VAC) Reservoir Capacity (L)
Availability Pricing
Julabo CORIO™ CP-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 1 kW Heater and 330 W at 20 °C Cooling, R290; 115 VAC, 60 Hz 1215326 Mfr # 9013713.N1.02
COMPARE
330 115 4
$14,761.89CAD / Each
Julabo CORIO™ CP-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 2 kW Heater and 330 W at 20 °C Cooling, R290; 200-230 VAC, 50/60 Hz 1215327 Mfr # 9013713.N1.33
COMPARE
330 200 / 230 4
$14,761.89CAD / Each
Julabo CORIO™ CP-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 1 kW Heater and 330 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz 1215328 Mfr # 9013713.S1.02
COMPARE
330 115 4
$14,614.56CAD / Each
Julabo CORIO™ CP-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 2 kW Heater and 330 W at 20 °C Cooling, R449A; 200-230 VAC, 50/60 Hz 1215329 Mfr # 9013713.S1.33
COMPARE
330 200 / 230 4
$14,614.56CAD / Each
Julabo CORIO™ CP-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 1 kW Heater and 440 W at 20 °C Cooling, R290; 115 VAC, 60 Hz 1215330 Mfr # 9013714.N1.02
COMPARE
440 115 4
$16,104.15CAD / Each
Julabo CORIO™ CP-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 2 kW Heater and 440 W at 20 °C Cooling, R290; 200-230 VAC, 50/60 Hz 1215331 Mfr # 9013714.N1.33
COMPARE
440 200 / 230 4
$16,104.15CAD / Each
Julabo CORIO™ CP-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 1 kW Heater and 440 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz 1215332 Mfr # 9013714.S1.02
COMPARE
440 115 4
$15,970.07CAD / Each
Julabo CORIO™ CP-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 200 °C, 2 kW Heater and 440 W at 20 °C Cooling, R449A; 200-230 VAC, 50/60 Hz 1215333 Mfr # 9013714.S1.33
COMPARE
440 200 / 230 4
$15,970.07CAD / Each
Julabo CORIO™ CP-200F Refrigerated/Heating Circulator, 4 L Tank, -20 to 200 °C, 1 kW Heater and 200 W at 20 °C Cooling, R134a; 115 VAC, 60 Hz 1215409 Mfr # 9013701.02
COMPARE
200 115 4
$13,496.25CAD / Each
Julabo CORIO™ CP-200F Refrigerated/Heating Circulator, 4 L Tank, -20 to 200 °C, 2 kW Heater and 200 W at 20 °C Cooling, R134a; 230 VAC, 50/60 Hz 1215410 Mfr # 9013701.33
COMPARE
200 230 4
$13,496.25CAD / Each
Julabo CORIO™ CP-300F Refrigerated/Heating Circulator, 4 L Tank, -25 to 200 °C, 1 kW Heater and 300 W at 20 °C Cooling, R134a; 115 VAC, 60 Hz 1215413 Mfr # 9013703.02
COMPARE
300 115 4
$15,115.51CAD / Each
Julabo CORIO™ CP-300F Refrigerated/Heating Circulator, 4 L Tank, -25 to 200 °C, 2 kW Heater and 300 W at 20 °C Cooling, R134a; 230 VAC, 50/60 Hz 1215414 Mfr # 9013703.33
COMPARE
300 230 4
$15,115.51CAD / Each
Julabo CORIO™CP-600F Refrigerated/Heating Circulator, 7.5 L Tank, -35 to 200 °C, 1 kW Heater and 600 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz 1215415 Mfr # 9013704.02
COMPARE
600 115 7.5
$18,107.96CAD / Each
Julabo CORIO™ CP-600F Refrigerated/Heating Circulator, 7.5 L Tank, -35 to 200 °C, 2 kW Heater and 600 W at 20 °C Cooling, R449A; 200-230 VAC, 50/60 Hz 1215416 Mfr # 9013704.33
COMPARE
600 200 / 230 7.5
$18,107.96CAD / Each
Julabo CORIO™ CP-1000F  Refrigerated/Heating Circulator, 7.5 L Tank, -50 to 200 °C, 1 kW Heater and 1000 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz 1215421 Mfr # 9013707.02
COMPARE
1000 115 7.5
$23,921.96CAD / Each
Julabo CORIO™ CP-1000F  Refrigerated/Heating Circulator, 7.5 L Tank, -50 to 200 °C, 2 kW Heater and 1000 W at 20 °C Cooling, R449A; 200-230 VAC, 50/60 Hz 1215422 Mfr # 9013707.33
COMPARE
1000 200 / 230 7.5
$23,921.96CAD / Each
Showing 1- 16 of 16