Imada Mechanical Force Gauges - Series FB

FEATURES
  • Simple, rugged, virtually maintenance free
  • Retains a high degree of accuracy for extended periods
  • Ergonomic, heavy-duty metal construction
  • No batteries needed
  • Intrinsically safe in hazardous environments
  • Capacity up to 100 lbs. (push and pull)
  • Overload capacity 165 lbs on all ranges
  • Real time and peak mode
  • Tare ring
  • Full set of accessories and carrying case included
  • Reverse display units are available
$633.56CAD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Imada FB-10 Analog Mechanical Force Gauge, 10 lb 9394974 Mfr # FB-10
COMPARE
Analog Mechanical Force Gauge, 10 lb
$633.56CAD / Each
Imada FB-100 Analog Mechanical Force Gauge, 100 lb 9394976 Mfr # FB-100
COMPARE
Analog Mechanical Force Gauge, 100 lb
$633.56CAD / Each
Imada FB-20 Analog Mechanical Force Gauge, 20 lb 9394982 Mfr # FB-20
COMPARE
Analog Mechanical Force Gauge, 20 lb
$633.56CAD / Each
Imada FB-30 Analog Mechanical Force Gauge, 30 lb 9394987 Mfr # FB-30
COMPARE
Analog Mechanical Force Gauge, 30 lb
$633.56CAD / Each
Imada FB-32OZ Analog Mechanical Force Gauge, 32 oz 9394991 Mfr # FB-32OZ
COMPARE
Analog Mechanical Force Gauge, 32 oz
$633.56CAD / Each
Imada FB-50 Analog Mechanical Force Gauge, 50 lb 9394994 Mfr # FB-50
COMPARE
Analog Mechanical Force Gauge, 50 lb
$633.56CAD / Each
Imada FB-500N Analog Mechanical Force Gauge, 500 N 9394995 Mfr # FB-500N
COMPARE
Analog Mechanical Force Gauge, 500 N
$633.56CAD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat