Imada FB-32OZ Analog Mechanical Force Gauge, 32 oz

Representative image only.

Imada FB-32OZ Analog Mechanical Force Gauge, 32 oz

Imada | Mfr: FB-32OZ| Item: 9394991

$633.56CAD / Each

for an additional
$278.10CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$633.56CAD / Each

Buy It Together

Current Item

Imada FB-32OZ Analog Mechanical Force Gauge, 32 oz
Imada FB-32OZ Analog Mechanical Force Gauge, 32 oz
9394991

$633.56CAD / Each
+ +
Imada OH-1 Optional Handle for Gauges
5985044

$163.91CAD / Each
=
Total Price 1075.57 ADD 3 ITEMS TO CART
Product Description

Key Features

  • 2 year warranty

More About this Item

FEATURES

  • Simple, rugged, virtually maintenance free
  • Retains a high degree of accuracy for extended periods
  • Ergonomic, heavy-duty metal construction
  • No batteries needed
  • Intrinsically safe in hazardous environments
  • Capacity up to 100 lbs. (push and pull)
  • Overload capacity 165 lbs on all ranges
  • Real time and peak mode
  • Tare ring
  • Full set of accessories and carrying case included
  • Reverse display units are available

Two year warranty
Specifications
  • Capacity (oz)32
  • Accuracy0.3% FS
  • DisplayAnalog dial with rotational tare ring
  • DescriptionAnalog Mechanical Force Gauge, 32 oz
  • Warranty2 year
Reviews
Q&A