IKA T50 ULTRA-TURRAX Heavy Duty Homogenizer
Choose a homogenizer based on your electrical requirements
  • Quickly homogenizes tissues and solids in batches from 0.25 to 30 liters
  • Integrated control ensures constant speed even with changes in viscosity
  • Output of 700 watts delivers superior dispersing power
The rotor-stator generator (sold separately) uses a pump-like action to constantly draw in sample. Through a combination of shear, impact, collision, and cavitation, the generator rapidly disperses and disintegrates the material suspended in the media. Homogenizer is designed for continuous operation and features an infinitely variable speed control from 500 to 10,000 rpm.
$7,629.21CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
IKA 3787001 T 50 ULTRA-TURRAX Heavy-Duty Homogenizer 115VAC-unit only 0473830 Mfr # 3787001
COMPARE
50/60 115
$7,629.21CAD / Each
IKA 3787000 T 50 ULTRA-TURRAX Heavy-Duty Homogenizer 230VAC-unit only 0473835 Mfr # 0003787000
COMPARE
50/60 220
$7,629.21CAD / Each