IDEX® PEEKsil™ Chromatography Tubing
Combines strength, consistency and biocompatibility
  • Fused silica covered by PEEK
  • Precut to standard lengths for convenience
In many analytical systems, PEEKsil tubing can be used as a direct replacement for conventional PEEK or stainless steel tubing. The fused silica core forms a rigid fluid pathway with extremely low adsorption, outstanding chemical compatibility, and extremely tight tolerances. The PEEK sheathing is very strong and seals well with many fitting types.
$81.77 - $327.09CAD / Pkg of 2
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (m) Tubing ID (mm) Tubing OD (in)
Availability Pricing
Idex 3255 Chromatography Tubing, PEEKsil™, 1/32" OD x 25 µm ID x 5 cm L, Orange; 2/PK 0201017 Mfr # 3255
COMPARE
0.05 0.025 1/32
$81.77CAD / Pkg of 2
Idex 3505 Chromatography Tubing, PEEKsil™, 1/32" OD x 50 µm ID x 5 cm L, Natural; 2/PK 0201021 Mfr # 3505
COMPARE
0.05 0.05 1/32
$88.11CAD / Pkg of 2
Idex 3755 Chromatography Tubing, PEEKsil™, 1/32" OD x 75 µm ID x 5 cm L, Black; 2/PK 0201178 Mfr # 3755
COMPARE
0.05 0.075 1/32
$88.11CAD / Pkg of 2
Idex 6255 Chromatography Tubing, PEEKsil™, 1/16" OD x 25 µm ID x 5 cm L, Orange; 5/PK 0201179 Mfr # 6255
COMPARE
0.05 0.025 5/8
$202.30CAD / Pkg of 5
Idex 6505 Chromatography Tubing, PEEKsil™, 1/16" OD x 50 µm ID x 5 cm L, Natural; 5/PK 0201180 Mfr # 6505
COMPARE
0.05 0.05 5/8
$203.03CAD / Pkg of 5
Idex 32515 Chromatography Tubing, PEEKsil™, 1/32" OD x 25 µm ID x 15 cm L, Orange; 2/PK 0201185 Mfr # 32515
COMPARE
0.15 0.025 1/32
$117.43CAD / Pkg of 2
Idex 32550 Chromatography Tubing, PEEKsil™, 1/32" OD x 25 µm ID x 50 cm L, Orange; 2/PK 0201187 Mfr # 32550
COMPARE
0.5 0.025 1/32
$216.74CAD / Pkg of 2
Idex 35010 Chromatography Tubing, PEEKsil™, 1/32" OD x 50 µm ID x 10 cm L, Natural; 2/PK 0201188 Mfr # 35010
COMPARE
0.1 0.05 1/32
$102.99CAD / Pkg of 2
Idex 35015 Chromatography Tubing, PEEKsil™, 1/32" OD x 50 µm ID x 15 cm L, Natural; 2/PK 0201189 Mfr # 35015
COMPARE
0.15 0.05 1/32
$110.80CAD / Pkg of 2
Idex 35020 Chromatography Tubing, PEEKsil™, 1/32" OD x 50 µm ID x 20 cm L, Natural; 2/PK 0201190 Mfr # 35020
COMPARE
0.2 0.05 1/32
$135.85CAD / Pkg of 2
Idex 35050 Chromatography Tubing, PEEKsil™, 1/32" OD x 50 µm ID x 50 cm L, Natural; 2/PK 0201191 Mfr # 35050
COMPARE
0.05 0.05 1/32
$214.08CAD / Pkg of 2
Idex 37520 Chromatography Tubing, PEEKsil™, 1/32" OD x 75 µm ID x 20 cm L, Black; 2/PK 0201194 Mfr # 37520
COMPARE
0.2 0.075 1/32
$126.42CAD / Pkg of 2
Idex 62510 Chromatography Tubing, PEEKsil™, 1/16" OD x 25 µm ID x 10 cm L, Orange; 5/PK 0201199 Mfr # 62510
COMPARE
0.1 0.025 5/8
$244.73CAD / Pkg of 5
Idex 62520 Chromatography Tubing, PEEKsil™, 1/16" OD x 25 µm ID x 20 cm L, Orange; 5/PK 0201205 Mfr # 62520
COMPARE
0.2 0.025 5/8
$302.04CAD / Pkg of 5
Idex 62550 Chromatography Tubing, PEEKsil™, 1/16" OD x 25 µm ID x 50 cm L, Orange; 2/PK 0201206 Mfr # 62550
COMPARE
0.5 0.025 5/8
$201.56CAD / Pkg of 2
Idex 63005 Chromatography Tubing, PEEKsil™, 1/16" OD x 300 µm ID x 5 cm L, Gray; 5/PK 0201207 Mfr # 63005
COMPARE
0.05 0.3 5/8
$209.07CAD / Pkg of 5
Idex 65010 Chromatography Tubing, PEEKsil™, 1/16" OD x 50 µm ID x 10 cm L, Natural; 5/PK 0201208 Mfr # 65010
COMPARE
0.1 0.05 5/8
$243.99CAD / Pkg of 5
Idex 65015 Chromatography Tubing, PEEKsil™, 1/16" OD x 50 µm ID x 15 cm L, Natural; 5/PK 0201209 Mfr # 65015
COMPARE
0.15 0.05 5/8
$261.82CAD / Pkg of 5
Idex 65050 Chromatography Tubing, PEEKsil™, 1/16" OD x 50 µm ID x 50 cm L, Natural; 2/PK 0201211 Mfr # 65050
COMPARE
0.5 0.05 5/8
$189.77CAD / Pkg of 2
Idex 67510 Chromatography Tubing, PEEKsil™, 1/16" OD x 75 µm ID x 10 cm L, Black; 5/PK 0201212 Mfr # 67510
COMPARE
0.1 0.075 5/8
$231.47CAD / Pkg of 5
Showing 1 - 20 of 33