IDEX® PEEK Tubing, 50 Ft Length

An excellent replacement for stainless steel tubing
  • Flexible PEEK tubing is biocompatible and chemically inert
  • Easily cut to desired lengths
  • Smooth internal surface decreases turbulence
  • Color-coded for easier identification
Applications: Chromatography
Characteristics: Unlike stainless steel, PEEK (polyetheretherketone) tubing is flexible and easy to cut. It provides excellent chemical resistance to both organic and inorganic solvents. Tubing withstands pressures to 5000 psi (3000 psi for model 02006-30). PEEK tubing has a very smooth internal surface, which causes less turbulence than metal tubing and improves resolution. Pack of 5-ft.
Certification: None
Maximum temperature: 212°F (100°C)
Sterilize: With autoclave, ethylene oxide, or gamma irradiation
$303.52 - $765.43CAD / Each

14 variations of this product are available.

See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tubing ID (in) Tubing OD (in)
Availability Pricing
Idex 1531BL Chromatography Tubing, Blue PEEK, 1/16" OD x 0.010" ID x 50 ft L; 1/EA 0200619 Mfr # 1531BL
COMPARE
Blue 1/100 1/16
$303.52CAD / Each
Idex 1531L Chromatography Tubing, Natural PEEK, 1/16" OD x 0.010" ID x 50 ft L; 1/EA 0200622 Mfr # 1531L
COMPARE
Natural 1/100 1/16
$303.52CAD / Each
Idex 1532L Chromatography Tubing, Orange PEEK, 1/16" OD x 0.020" ID x 50 ft L; 1/EA 0200625 Mfr # 1532L
COMPARE
Orange 1/50 1/16
$303.52CAD / Each
Idex 1533L Chromatography Tubing, Green PEEK, 1/16" OD x 0.030" ID x 50 ft L; 1/EA 0200627 Mfr # 1533L
COMPARE
Green 1/32 1/16
$303.52CAD / Each
Idex 1535L Chromatography Tubing, Red PEEK, 1/16" OD x 0.005" ID x 50 ft L; 1/EA 0200631 Mfr # 1535L
COMPARE
Red 1/200 1/16
$436.12CAD / Each
Idex 1536L Chromatography Tubing, Yellow PEEK, 1/16" OD x 0.007" ID x 50 ft L; 1/EA 0200633 Mfr # 1536L
COMPARE
Yellow 1/150 1/16
$303.52CAD / Each
Idex 1538L Chromatography Tubing, Natural PEEK, 1/16" OD x 0.040" ID x 50 ft L; 1/EA 0200635 Mfr # 1538L
COMPARE
Natural 1/25 1/16
$303.52CAD / Each
Idex 1560L Chromatography Tubing, Natural PEEK, 1/16" OD x 0.0025" ID x 50 ft L; 1/EA 0200638 Mfr # 1560L
COMPARE
Natural 1/500 1/16
$448.65CAD / Each
Idex 1561L Chromatography Tubing, Black PEEK, 1/16" OD x 0.004" ID x 50 ft L; 1/EA 0200640 Mfr # 1561L
COMPARE
Black 1/250 1/16
$431.70CAD / Each
Idex 1568L Chromatography Tubing, Natural PEEK, 1/32" OD x 0.015" ID x 50 ft L; 1/EA 0200642 Mfr # 1568L
COMPARE
Natural 1/64 1/32
$515.69CAD / Each
Idex 1569L Chromatography Tubing, Orange PEEK, 1/32" OD x 0.020" ID x 50 ft L; 1/EA 0200644 Mfr # 1569L
COMPARE
Orange 1/50 1/32
$765.43CAD / Each
Idex 1576L Chromatography Tubing, Red PEEK, 1/32" OD x 0.005" ID x 50 ft L; 1/EA 0200646 Mfr # 1576L
COMPARE
Red 1/200 1/32
$515.69CAD / Each
Idex 1577L Chromatography Tubing, Yellow PEEK, 1/32" OD x 0.007" ID x 50 ft L; 1/EA 0200648 Mfr # 1577L
COMPARE
Yellow 1/150 1/32
$765.43CAD / Each
Idex Chromatography Tubing, Blue PEEK, 1/32" OD x 0.010" ID x 50 ft L 0200651 Mfr # 1581L
COMPARE
Blue 1/100 1/32
$515.69CAD / Each
Showing 1- 14 of 14

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat